Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 67505079-67505388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPC
Entrez Gene ID
1060
Gene Name
centromere protein C
Gene Aliases
CENP-C, CENPC1, MIF2, hcp-4
UniGene
Hs.479867
Gene Chromosome Location
Chr.4: 67468762-67545601 on Build GRCh38
Transcripts
NM_001812.2 XM_006714064.2 XM_011531542.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3303813 c.2120A>G p.H707R Chr.4: 67505216-67505216 on Build GRCh38
6567407 c.2052-6C>G p.? Chr.4: 67505290-67505290 on Build GRCh38
1430584 c.2107G>A p.D703N Chr.4: 67505229-67505229 on Build GRCh38
5356049 c.2077T>A p.Y693N Chr.4: 67505259-67505259 on Build GRCh38
4534161 c.2054G>A p.G685E Chr.4: 67505282-67505282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778586358 Chr.4: 67505210-67505210 on Build GRCh38
rs753436526 Chr.4: 67505198-67505198 on Build GRCh38
rs1014766861 Chr.4: 67505134-67505134 on Build GRCh38
rs913411048 Chr.4: 67505280-67505280 on Build GRCh38
rs768653284 Chr.4: 67505286-67505286 on Build GRCh38
rs773594453 Chr.4: 67505157-67505157 on Build GRCh38
rs909088764 Chr.4: 67505355-67505355 on Build GRCh38
rs758212108 Chr.4: 67505225-67505225 on Build GRCh38
rs1029038060 Chr.4: 67505306-67505306 on Build GRCh38
rs761146385 Chr.4: 67505330-67505330 on Build GRCh38
rs1022651302 Chr.4: 67505229-67505229 on Build GRCh38
rs965797098 Chr.4: 67505281-67505281 on Build GRCh38
rs767708043 Chr.4: 67505127-67505127 on Build GRCh38
rs1036607076 Chr.4: 67505128-67505128 on Build GRCh38
rs987563130 Chr.4: 67505322-67505322 on Build GRCh38
rs746939194 Chr.4: 67505251-67505251 on Build GRCh38
rs774572311 Chr.4: 67505290-67505290 on Build GRCh38
rs1033258529 Chr.4: 67505254-67505254 on Build GRCh38
rs189594633 Chr.4: 67505272-67505272 on Build GRCh38
rs201011714 Chr.4: 67505259-67505259 on Build GRCh38
rs777769961 Chr.4: 67505238-67505238 on Build GRCh38
rs927330768 Chr.4: 67505269-67505269 on Build GRCh38
rs767860654 Chr.4: 67505313-67505313 on Build GRCh38
rs375275429 Chr.4: 67505196-67505196 on Build GRCh38
rs371536161 Chr.4: 67505193-67505193 on Build GRCh38
rs772693518 Chr.4: 67505185-67505186 on Build GRCh38
rs367862812 Chr.4: 67505204-67505204 on Build GRCh38
rs375531779 Chr.4: 67505317-67505317 on Build GRCh38
rs1034806944 Chr.4: 67505140-67505140 on Build GRCh38
rs921592405 Chr.4: 67505302-67505302 on Build GRCh38
rs749342568 Chr.4: 67505282-67505282 on Build GRCh38
rs912026390 Chr.4: 67505361-67505361 on Build GRCh38
rs747761733 Chr.4: 67505216-67505216 on Build GRCh38
rs759651198 Chr.4: 67505174-67505175 on Build GRCh38
rs904931312 Chr.4: 67505113-67505113 on Build GRCh38
rs759979935 Chr.4: 67505194-67505194 on Build GRCh38
rs1003284013 Chr.4: 67505133-67505133 on Build GRCh38
rs761995385 Chr.4: 67505310-67505310 on Build GRCh38
rs201792278 Chr.4: 67505187-67505187 on Build GRCh38
rs750912850 Chr.4: 67505183-67505183 on Build GRCh38
rs766822126 Chr.4: 67505331-67505331 on Build GRCh38

View Full Product Details