Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173159874-173160227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIP1
Entrez Gene ID
662
Gene Name
BCL2 interacting protein 1
Gene Aliases
NIP1, SEC20, TRG-8
UniGene
Hs.145726
Gene Chromosome Location
Chr.5: 173144442-173164387 on Build GRCh38
Transcripts
NM_001205.2 NM_013978.2 NM_013979.2 NM_013980.2 XM_011534638.1 XM_011534639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6612182 c.540C>T p.T180T Chr.5: 173159972-173159972 on Build GRCh38
1730759 c.609G>T p.M203I Chr.5: 173160041-173160041 on Build GRCh38
5403095 c.588C>T p.V196V Chr.5: 173160020-173160020 on Build GRCh38
1191029 c.561_562insG p.I188fs*31 Chr.5: 173159993-173159994 on Build GRCh38
3975415 c.570G>A p.R190R Chr.5: 173160002-173160002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940780544 Chr.5: 173160009-173160009 on Build GRCh38
rs776688323 Chr.5: 173160032-173160032 on Build GRCh38
rs767436623 Chr.5: 173159963-173159963 on Build GRCh38
rs928032803 Chr.5: 173160005-173160005 on Build GRCh38
rs370701128 Chr.5: 173160072-173160072 on Build GRCh38
rs1032821854 Chr.5: 173160150-173160150 on Build GRCh38
rs982602564 Chr.5: 173159991-173159991 on Build GRCh38
rs930865432 Chr.5: 173160058-173160058 on Build GRCh38
rs969394774 Chr.5: 173159971-173159971 on Build GRCh38
rs200645088 Chr.5: 173160011-173160011 on Build GRCh38
rs181667978 Chr.5: 173160090-173160090 on Build GRCh38
rs555189519 Chr.5: 173159909-173159909 on Build GRCh38
rs559764497 Chr.5: 173159905-173159905 on Build GRCh38
rs921920126 Chr.5: 173160151-173160151 on Build GRCh38
rs994556157 Chr.5: 173159957-173159957 on Build GRCh38
rs144912663 Chr.5: 173160017-173160017 on Build GRCh38
rs752014975 Chr.5: 173159987-173159987 on Build GRCh38
rs761908319 Chr.5: 173160042-173160042 on Build GRCh38
rs778219066 Chr.5: 173159985-173159985 on Build GRCh38
rs761669245 Chr.5: 173159910-173159910 on Build GRCh38
rs991383052 Chr.5: 173159940-173159940 on Build GRCh38
rs746561482 Chr.5: 173160029-173160029 on Build GRCh38
rs974977954 Chr.5: 173160146-173160146 on Build GRCh38
rs768091546 Chr.5: 173160030-173160030 on Build GRCh38
rs563673695 Chr.5: 173160077-173160077 on Build GRCh38
rs773152412 Chr.5: 173160062-173160062 on Build GRCh38
rs545317197 Chr.5: 173160021-173160021 on Build GRCh38
rs745449207 Chr.5: 173160023-173160023 on Build GRCh38
rs930100640 Chr.5: 173159993-173159993 on Build GRCh38
rs769645693 Chr.5: 173160054-173160054 on Build GRCh38
rs752841614 Chr.5: 173159964-173159964 on Build GRCh38
rs760473355 Chr.5: 173159895-173159895 on Build GRCh38
rs759627037 Chr.5: 173160069-173160069 on Build GRCh38
rs902897029 Chr.5: 173160132-173160132 on Build GRCh38
rs966242876 Chr.5: 173160103-173160103 on Build GRCh38
rs768662885 Chr.5: 173159899-173159899 on Build GRCh38
rs1019095412 Chr.5: 173160091-173160091 on Build GRCh38
rs779321350 Chr.5: 173160019-173160019 on Build GRCh38
rs944392718 Chr.5: 173160120-173160120 on Build GRCh38
rs751826984 Chr.5: 173159929-173159929 on Build GRCh38
rs754388187 Chr.5: 173160002-173160002 on Build GRCh38
rs543894811 Chr.5: 173160027-173160027 on Build GRCh38
rs759807894 Chr.5: 173159951-173159951 on Build GRCh38
rs187068256 Chr.5: 173160186-173160186 on Build GRCh38
rs754343300 Chr.5: 173160097-173160097 on Build GRCh38
rs370658787 Chr.5: 173159967-173159967 on Build GRCh38
rs764220856 Chr.5: 173160092-173160092 on Build GRCh38
rs767808313 Chr.5: 173160070-173160070 on Build GRCh38
rs779385258 Chr.5: 173160028-173160028 on Build GRCh38
rs377348075 Chr.5: 173159918-173159918 on Build GRCh38
rs1040445754 Chr.5: 173160121-173160121 on Build GRCh38
rs757642101 Chr.5: 173160061-173160065 on Build GRCh38

View Full Product Details