Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 39086563-39087022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP3
Entrez Gene ID
79937
Gene Name
contactin associated protein-like 3
Gene Aliases
CASPR3, CNTNAP3A
UniGene
Hs.128474
Gene Chromosome Location
Chr.9: 39072767-39288303 on Build GRCh38
Transcripts
NM_033655.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3982867 c.3108A>G p.Q1036Q Chr.9: 39086698-39086698 on Build GRCh38
6115889 c.2989C>T p.H997Y Chr.9: 39086817-39086817 on Build GRCh38
6291188 c.3291C>T p.N1097N Chr.9: 39086779-39086779 on Build GRCh38
3930074 c.2976C>G p.Y992* Chr.9: 39086830-39086830 on Build GRCh38
1109158 c.3312C>A p.H1104Q Chr.9: 39086758-39086758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749621100 Chr.9: 39086852-39086852 on Build GRCh38
rs1002762297 Chr.9: 39086966-39086966 on Build GRCh38
rs896725221 Chr.9: 39086994-39086994 on Build GRCh38
rs749204525 Chr.9: 39086694-39086694 on Build GRCh38
rs2183892 Chr.9: 39086698-39086698 on Build GRCh38
rs777300631 Chr.9: 39086830-39086830 on Build GRCh38
rs762370069 Chr.9: 39086775-39086775 on Build GRCh38
rs780302198 Chr.9: 39086682-39086685 on Build GRCh38
rs573860982 Chr.9: 39086780-39086780 on Build GRCh38
rs1056671340 Chr.9: 39086975-39086975 on Build GRCh38
rs539815325 Chr.9: 39086663-39086663 on Build GRCh38
rs905305277 Chr.9: 39086962-39086962 on Build GRCh38
rs988207216 Chr.9: 39086642-39086642 on Build GRCh38
rs756299748 Chr.9: 39086672-39086674 on Build GRCh38
rs752663189 Chr.9: 39086646-39086646 on Build GRCh38
rs775778547 Chr.9: 39086739-39086739 on Build GRCh38
rs182537264 Chr.9: 39086779-39086779 on Build GRCh38
rs764148892 Chr.9: 39086654-39086654 on Build GRCh38
rs373340917 Chr.9: 39086844-39086844 on Build GRCh38
rs2778040 Chr.9: 39086999-39086999 on Build GRCh38
rs941359071 Chr.9: 39086907-39086907 on Build GRCh38
rs765839854 Chr.9: 39086777-39086777 on Build GRCh38
rs10814793 Chr.9: 39086943-39086943 on Build GRCh38
rs2778041 Chr.9: 39086669-39086669 on Build GRCh38
rs759339531 Chr.9: 39086732-39086732 on Build GRCh38
rs754606597 Chr.9: 39086675-39086675 on Build GRCh38
rs755747596 Chr.9: 39086807-39086807 on Build GRCh38
rs537352145 Chr.9: 39086634-39086635 on Build GRCh38
rs943168816 Chr.9: 39086741-39086741 on Build GRCh38
rs3071057 Chr.9: 39086640-39086641 on Build GRCh38
rs780347143 Chr.9: 39086796-39086796 on Build GRCh38
rs2721734 Chr.9: 39086731-39086731 on Build GRCh38
rs929279212 Chr.9: 39086823-39086823 on Build GRCh38
rs557261055 Chr.9: 39086648-39086648 on Build GRCh38
rs559505561 Chr.9: 39086756-39086758 on Build GRCh38
rs569184734 Chr.9: 39086660-39086660 on Build GRCh38
rs752375430 Chr.9: 39086803-39086803 on Build GRCh38
rs760848518 Chr.9: 39086747-39086747 on Build GRCh38
rs371742242 Chr.9: 39086638-39086639 on Build GRCh38
rs780907623 Chr.9: 39086679-39086679 on Build GRCh38
rs979113279 Chr.9: 39086715-39086716 on Build GRCh38
rs540321225 Chr.9: 39086695-39086695 on Build GRCh38
rs562821159 Chr.9: 39086932-39086932 on Build GRCh38
rs779371415 Chr.9: 39086664-39086664 on Build GRCh38
rs750647230 Chr.9: 39086636-39086637 on Build GRCh38
rs767731653 Chr.9: 39086640-39086640 on Build GRCh38
rs146610751 Chr.9: 39086758-39086758 on Build GRCh38
rs770797219 Chr.9: 39086853-39086853 on Build GRCh38
rs191597258 Chr.9: 39086680-39086680 on Build GRCh38
rs777199474 Chr.9: 39086691-39086691 on Build GRCh38
rs754337641 Chr.9: 39086655-39086655 on Build GRCh38
rs753885289 Chr.9: 39086768-39086768 on Build GRCh38
rs778810050 Chr.9: 39086863-39086863 on Build GRCh38
rs144374834 Chr.9: 39086735-39086735 on Build GRCh38
rs267602246 Chr.9: 39086808-39086808 on Build GRCh38
rs202227389 Chr.9: 39086722-39086722 on Build GRCh38
rs200008891 Chr.9: 39086690-39086690 on Build GRCh38

View Full Product Details