Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36767914-36768225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSN1
Entrez Gene ID
79980
Gene Name
DSN1 homolog, MIS12 kinetochore complex component
Gene Aliases
C20orf172, KNL3, MIS13, dJ469A13.2, hKNL-3
UniGene
Hs.632268
Gene Chromosome Location
Chr.20: 36751791-36773827 on Build GRCh38
Transcripts
NM_001145315.1 NM_001145316.1 NM_001145317.1 NM_001145318.1 NM_024918.3 XM_006723876.2 XM_017028070.1 XM_017028071.1 XM_017028072.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
234227 c.427C>T p.Q143* Chr.20: 36767971-36767971 on Build GRCh38
7143147 c.360C>G p.L120L Chr.20: 36768038-36768038 on Build GRCh38
6649644 c.397C>T p.R133W Chr.20: 36768001-36768001 on Build GRCh38
6214490 c.358C>G p.L120V Chr.20: 36768040-36768040 on Build GRCh38
3545890 c.366G>A p.R122R Chr.20: 36768032-36768032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763643000 Chr.20: 36768070-36768070 on Build GRCh38
rs536992930 Chr.20: 36768113-36768113 on Build GRCh38
rs770135730 Chr.20: 36768040-36768040 on Build GRCh38
rs74470097 Chr.20: 36767995-36767995 on Build GRCh38
rs534146241 Chr.20: 36768078-36768078 on Build GRCh38
rs1026572167 Chr.20: 36767964-36767964 on Build GRCh38
rs995145772 Chr.20: 36767948-36767948 on Build GRCh38
rs750266501 Chr.20: 36768086-36768086 on Build GRCh38
rs780232179 Chr.20: 36768033-36768033 on Build GRCh38
rs750994643 Chr.20: 36767950-36767950 on Build GRCh38
rs745734102 Chr.20: 36768020-36768020 on Build GRCh38
rs766924419 Chr.20: 36768046-36768046 on Build GRCh38
rs755746357 Chr.20: 36767971-36767971 on Build GRCh38
rs757135566 Chr.20: 36768000-36768000 on Build GRCh38
rs763460716 Chr.20: 36767956-36767956 on Build GRCh38
rs752230934 Chr.20: 36767959-36767959 on Build GRCh38
rs372066834 Chr.20: 36768034-36768034 on Build GRCh38
rs932595867 Chr.20: 36768147-36768147 on Build GRCh38
rs368233235 Chr.20: 36768001-36768001 on Build GRCh38
rs921891370 Chr.20: 36768115-36768115 on Build GRCh38
rs572104227 Chr.20: 36768095-36768095 on Build GRCh38
rs926094102 Chr.20: 36768188-36768188 on Build GRCh38
rs763405668 Chr.20: 36768045-36768045 on Build GRCh38
rs773567248 Chr.20: 36768043-36768043 on Build GRCh38
rs774954008 Chr.20: 36768047-36768047 on Build GRCh38
rs777524503 Chr.20: 36767973-36767973 on Build GRCh38
rs759546323 Chr.20: 36768100-36768100 on Build GRCh38
rs375360494 Chr.20: 36768056-36768056 on Build GRCh38
rs553777811 Chr.20: 36768076-36768076 on Build GRCh38
rs758441579 Chr.20: 36768025-36768025 on Build GRCh38
rs746698351 Chr.20: 36768028-36768028 on Build GRCh38
rs571380182 Chr.20: 36768037-36768037 on Build GRCh38
rs977688972 Chr.20: 36768140-36768140 on Build GRCh38
rs753573338 Chr.20: 36767977-36767977 on Build GRCh38

View Full Product Details