Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152329706-152329866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW7
Entrez Gene ID
55294
Gene Name
F-box and WD repeat domain containing 7
Gene Aliases
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
UniGene
Hs.561245
Gene Chromosome Location
Chr.4: 152321258-152536095 on Build GRCh38
Transcripts
NM_001013415.1 NM_018315.4 NM_033632.3 XM_011532083.1 XM_011532084.1 XM_011532085.1 XM_011532086.1 XM_011532087.1 XM_011532088.1 XM_017008362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7430541 c.1195G>T p.D399Y Chr.4: 152329713-152329713 on Build GRCh38
6978105 c.790_792delCAT p.H264delH Chr.4: 152329762-152329764 on Build GRCh38
1309815 c.800C>A p.T267K Chr.4: 152329754-152329754 on Build GRCh38
1052104 c.460C>T p.R154* Chr.4: 152329731-152329731 on Build GRCh38
6967304 c.476C>A p.S159Y Chr.4: 152329715-152329715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187946218 Chr.4: 152329813-152329813 on Build GRCh38
rs751504450 Chr.4: 152329796-152329796 on Build GRCh38
rs183836512 Chr.4: 152329808-152329808 on Build GRCh38
rs754942103 Chr.4: 152329797-152329797 on Build GRCh38
rs756345085 Chr.4: 152329747-152329747 on Build GRCh38
rs760083752 Chr.4: 152329830-152329830 on Build GRCh38
rs747552028 Chr.4: 152329815-152329815 on Build GRCh38
rs904144155 Chr.4: 152329789-152329789 on Build GRCh38
rs757351274 Chr.4: 152329788-152329788 on Build GRCh38
rs372149288 Chr.4: 152329827-152329827 on Build GRCh38
rs936906897 Chr.4: 152329799-152329799 on Build GRCh38
rs781030534 Chr.4: 152329804-152329804 on Build GRCh38
rs772511577 Chr.4: 152329831-152329831 on Build GRCh38
rs370526423 Chr.4: 152329811-152329811 on Build GRCh38
rs866269762 Chr.4: 152329754-152329754 on Build GRCh38
rs775893298 Chr.4: 152329836-152329836 on Build GRCh38
rs779912228 Chr.4: 152329790-152329790 on Build GRCh38
rs753876467 Chr.4: 152329777-152329777 on Build GRCh38
rs767619725 Chr.4: 152329812-152329813 on Build GRCh38
rs774930781 Chr.4: 152329829-152329829 on Build GRCh38
rs377257045 Chr.4: 152329765-152329765 on Build GRCh38
rs747938212 Chr.4: 152329807-152329807 on Build GRCh38
rs57047792 Chr.4: 152329782-152329782 on Build GRCh38
rs770577602 Chr.4: 152329821-152329821 on Build GRCh38

View Full Product Details