Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 393228-393354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF4
Entrez Gene ID
3662
Gene Name
interferon regulatory factor 4
Gene Aliases
LSIRF, MUM1, NF-EM5, SHEP8
UniGene
Hs.401013
Gene Chromosome Location
Chr.6: 391739-411443 on Build GRCh38
Transcripts
NM_001195286.1 NM_002460.3 NR_046000.2 XM_006715090.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6930637 c.81G>A p.W27* Chr.6: 393233-393233 on Build GRCh38
5949955 c.166C>T p.H56Y Chr.6: 393318-393318 on Build GRCh38
7424247 c.103G>T p.G35C Chr.6: 393255-393255 on Build GRCh38
5947065 c.174C>T p.G58G Chr.6: 393326-393326 on Build GRCh38
220752 c.144C>A p.S48R Chr.6: 393296-393296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991845330 Chr.6: 393299-393299 on Build GRCh38
rs748194946 Chr.6: 393317-393317 on Build GRCh38
rs753781892 Chr.6: 393285-393287 on Build GRCh38
rs781441118 Chr.6: 393310-393310 on Build GRCh38
rs764159153 Chr.6: 393281-393281 on Build GRCh38
rs538907751 Chr.6: 393256-393256 on Build GRCh38
rs747212033 Chr.6: 393302-393302 on Build GRCh38
rs755175509 Chr.6: 393308-393308 on Build GRCh38
rs751037944 Chr.6: 393273-393273 on Build GRCh38
rs934099606 Chr.6: 393285-393285 on Build GRCh38
rs755738102 Chr.6: 393265-393265 on Build GRCh38
rs780297970 Chr.6: 393288-393288 on Build GRCh38
rs144020240 Chr.6: 393272-393272 on Build GRCh38
rs114321276 Chr.6: 393323-393323 on Build GRCh38
rs369688140 Chr.6: 393264-393264 on Build GRCh38

View Full Product Details