Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152324317-152324475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW7
Entrez Gene ID
55294
Gene Name
F-box and WD repeat domain containing 7
Gene Aliases
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
UniGene
Hs.561245
Gene Chromosome Location
Chr.4: 152321258-152536095 on Build GRCh38
Transcripts
NM_001013415.1 NM_018315.4 NM_033632.3 XM_011532083.1 XM_011532084.1 XM_011532085.1 XM_011532086.1 XM_011532087.1 XM_011532088.1 XM_017008362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
222957 c.1685C>T p.S562L Chr.4: 152324354-152324354 on Build GRCh38
6197910 c.1645-4G>A p.? Chr.4: 152324398-152324398 on Build GRCh38
7339157 c.1694T>G p.V565G Chr.4: 152324345-152324345 on Build GRCh38
6191361 c.1698G>A p.W566* Chr.4: 152324341-152324341 on Build GRCh38
4122756 c.1323T>C p.L441L Chr.4: 152324362-152324362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142961063 Chr.4: 152324380-152324380 on Build GRCh38
rs752635282 Chr.4: 152324414-152324414 on Build GRCh38
rs1024060344 Chr.4: 152324349-152324349 on Build GRCh38
rs749772265 Chr.4: 152324398-152324398 on Build GRCh38
rs752171411 Chr.4: 152324356-152324356 on Build GRCh38
rs779134112 Chr.4: 152324409-152324409 on Build GRCh38
rs936096831 Chr.4: 152324428-152324428 on Build GRCh38
rs1001597080 Chr.4: 152324387-152324387 on Build GRCh38
rs757851336 Chr.4: 152324405-152324405 on Build GRCh38
rs746293581 Chr.4: 152324443-152324443 on Build GRCh38
rs138665286 Chr.4: 152324347-152324347 on Build GRCh38
rs962968551 Chr.4: 152324434-152324434 on Build GRCh38
rs755652430 Chr.4: 152324368-152324368 on Build GRCh38
rs375373188 Chr.4: 152324446-152324446 on Build GRCh38
rs371077307 Chr.4: 152324420-152324420 on Build GRCh38

View Full Product Details