Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25243846-25244004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3A
Entrez Gene ID
1788
Gene Name
DNA methyltransferase 3 alpha
Gene Aliases
DNMT3A2, M.HsaIIIA, TBRS
UniGene
Hs.515840
Gene Chromosome Location
Chr.2: 25232961-25342590 on Build GRCh38
Transcripts
NM_001320893.1 NM_022552.4 NM_153759.3 NM_175629.2 NR_135490.1 XM_005264175.4 XM_005264177.4 XM_011532662.2 XM_011532663.2 XM_011532664.2 XM_011532666.2 XM_011532667.2 XM_017003526.1 XM_017003527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
231550 c.1904G>C p.R635P Chr.2: 25243930-25243930 on Build GRCh38
7184714 c.1897C>T p.P633S Chr.2: 25243937-25243937 on Build GRCh38
6945536 c.1887G>T p.E629D Chr.2: 25243947-25243947 on Build GRCh38
133741 c.1865_1866insGT p.Y623fs*29 Chr.2: 25243968-25243969 on Build GRCh38
6498437 c.1936+8G>A p.? Chr.2: 25243890-25243890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763147762 Chr.2: 25243926-25243926 on Build GRCh38
rs144689354 Chr.2: 25243931-25243931 on Build GRCh38
rs888799927 Chr.2: 25243873-25243873 on Build GRCh38
rs775516996 Chr.2: 25243867-25243867 on Build GRCh38
rs781503197 Chr.2: 25243980-25243980 on Build GRCh38
rs948196657 Chr.2: 25243868-25243868 on Build GRCh38
rs906581890 Chr.2: 25243905-25243905 on Build GRCh38
rs929806900 Chr.2: 25243927-25243927 on Build GRCh38
rs745381125 Chr.2: 25243962-25243962 on Build GRCh38
rs529573762 Chr.2: 25243890-25243890 on Build GRCh38
rs1017971955 Chr.2: 25243974-25243974 on Build GRCh38
rs751562376 Chr.2: 25243930-25243930 on Build GRCh38
rs368148016 Chr.2: 25243865-25243865 on Build GRCh38
rs371523368 Chr.2: 25243924-25243924 on Build GRCh38
rs778979256 Chr.2: 25243928-25243928 on Build GRCh38
rs551213321 Chr.2: 25243910-25243910 on Build GRCh38
rs866791048 Chr.2: 25243983-25243983 on Build GRCh38
rs771876830 Chr.2: 25243866-25243866 on Build GRCh38
rs1046919792 Chr.2: 25243891-25243891 on Build GRCh38
rs376550450 Chr.2: 25243928-25243928 on Build GRCh38
rs149876809 Chr.2: 25243951-25243951 on Build GRCh38

View Full Product Details