Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108310212-108310375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATM
Entrez Gene ID
472
Gene Name
ATM serine/threonine kinase
Gene Aliases
AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
UniGene
Hs.367437
Gene Chromosome Location
Chr.11: 108222484-108369102 on Build GRCh38
Transcripts
NM_000051.3 XM_005271561.4 XM_005271562.4 XM_006718843.3 XM_006718845.1 XM_011542840.2 XM_011542842.2 XM_011542843.2 XM_011542844.2 XM_011542845.1 XM_017017789.1 XM_017017790.1 XM_017017791.1 XM_017017249.1 XM_017017250.1

Gene Symbol
C11orf65
Entrez Gene ID
160140
Gene Name
chromosome 11 open reading frame 65
Gene Aliases
-
UniGene
Hs.653180
Gene Chromosome Location
Chr.11: 108307344-108469832 on Build GRCh38
Transcripts
NM_000051.3 XM_005271561.4 XM_005271562.4 XM_006718843.3 XM_006718845.1 XM_011542840.2 XM_011542842.2 XM_011542843.2 XM_011542844.2 XM_011542845.1 XM_017017789.1 XM_017017790.1 XM_017017791.1 XM_017017249.1 XM_017017250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6906457 c.5849C>A p.A1950D Chr.11: 108310246-108310246 on Build GRCh38
922730 c.5834_5835delCT p.Q1946fs*18 Chr.11: 108310231-108310232 on Build GRCh38
6956867 c.5892G>C p.K1964N Chr.11: 108310289-108310289 on Build GRCh38
922729 c.5823T>C p.V1941V Chr.11: 108310220-108310220 on Build GRCh38
5177086 c.5914delA p.R1973fs*17 Chr.11: 108310311-108310311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776150043 Chr.11: 108310241-108310241 on Build GRCh38
rs587782503 Chr.11: 108310276-108310276 on Build GRCh38
rs864622148 Chr.11: 108310284-108310284 on Build GRCh38
rs3092911 Chr.11: 108310331-108310331 on Build GRCh38
rs786202089 Chr.11: 108310314-108310314 on Build GRCh38
rs730881377 Chr.11: 108310312-108310312 on Build GRCh38
rs876658989 Chr.11: 108310264-108310264 on Build GRCh38
rs376169886 Chr.11: 108310250-108310250 on Build GRCh38
rs876660515 Chr.11: 108310273-108310273 on Build GRCh38
rs587781722 Chr.11: 108310305-108310305 on Build GRCh38
rs201963507 Chr.11: 108310287-108310287 on Build GRCh38
rs1046092637 Chr.11: 108310295-108310295 on Build GRCh38
rs56399311 Chr.11: 108310279-108310279 on Build GRCh38
rs786202728 Chr.11: 108310289-108310289 on Build GRCh38
rs368629884 Chr.11: 108310337-108310337 on Build GRCh38
rs864622297 Chr.11: 108310253-108310253 on Build GRCh38
rs540054724 Chr.11: 108310265-108310265 on Build GRCh38
rs587781963 Chr.11: 108310255-108310255 on Build GRCh38
rs587782198 Chr.11: 108310307-108310307 on Build GRCh38
rs587781727 Chr.11: 108310290-108310294 on Build GRCh38
rs751786087 Chr.11: 108310345-108310345 on Build GRCh38
rs759873743 Chr.11: 108310347-108310347 on Build GRCh38

View Full Product Details