Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 401347-401474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF4
Entrez Gene ID
3662
Gene Name
interferon regulatory factor 4
Gene Aliases
LSIRF, MUM1, NF-EM5, SHEP8
UniGene
Hs.401013
Gene Chromosome Location
Chr.6: 391739-411443 on Build GRCh38
Transcripts
NM_001195286.1 NM_002460.3 NR_046000.2 XM_006715090.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
741980 c.758A>C p.H253P Chr.6: 401436-401436 on Build GRCh38
5607445 c.778G>A p.E260K Chr.6: 401456-401456 on Build GRCh38
6957217 c.764G>A p.C255Y Chr.6: 401442-401442 on Build GRCh38
3627068 c.779A>C p.E260A Chr.6: 401457-401457 on Build GRCh38
4390933 c.746-1G>A p.? Chr.6: 401423-401423 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs148760922 Chr.6: 401444-401444 on Build GRCh38
rs375273531 Chr.6: 401379-401379 on Build GRCh38
rs1045720105 Chr.6: 401372-401372 on Build GRCh38
rs765592332 Chr.6: 401405-401405 on Build GRCh38
rs376012784 Chr.6: 401406-401406 on Build GRCh38
rs369116605 Chr.6: 401389-401389 on Build GRCh38
rs199834880 Chr.6: 401430-401430 on Build GRCh38
rs781276700 Chr.6: 401454-401454 on Build GRCh38
rs758839744 Chr.6: 401425-401425 on Build GRCh38
rs912246620 Chr.6: 401434-401434 on Build GRCh38
rs115625962 Chr.6: 401446-401446 on Build GRCh38
rs975308028 Chr.6: 401448-401448 on Build GRCh38
rs897889268 Chr.6: 401412-401412 on Build GRCh38
rs774119356 Chr.6: 401377-401377 on Build GRCh38
rs759801296 Chr.6: 401376-401376 on Build GRCh38
rs780137774 Chr.6: 401431-401431 on Build GRCh38
rs564409022 Chr.6: 401392-401392 on Build GRCh38
rs939320685 Chr.6: 401384-401384 on Build GRCh38

View Full Product Details