Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 112767216-112767346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APC
Entrez Gene ID
324
Gene Name
APC, WNT signaling pathway regulator
Gene Aliases
BTPS2, DP2, DP2.5, DP3, GS, PPP1R46
UniGene
Hs.158932
Gene Chromosome Location
Chr.5: 112707505-112846239 on Build GRCh38
Transcripts
NM_000038.5 NM_001127510.2 NM_001127511.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5825016 c.275C>G p.S92C Chr.5: 112767243-112767243 on Build GRCh38
1696038 c.284C>T p.S95F Chr.5: 112767252-112767252 on Build GRCh38
6964915 c.317delG p.R106fs*19 Chr.5: 112767285-112767285 on Build GRCh38
4486585 c.276C>T p.S92S Chr.5: 112767244-112767244 on Build GRCh38
48234 c.350C>T p.S117L Chr.5: 112767318-112767318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200117298 Chr.5: 112767309-112767309 on Build GRCh38
rs886059793 Chr.5: 112767304-112767304 on Build GRCh38
rs776242276 Chr.5: 112767281-112767281 on Build GRCh38
rs769708176 Chr.5: 112767243-112767243 on Build GRCh38
rs376213437 Chr.5: 112767256-112767256 on Build GRCh38
rs774229223 Chr.5: 112767249-112767249 on Build GRCh38
rs786202322 Chr.5: 112767300-112767300 on Build GRCh38
rs201567345 Chr.5: 112767245-112767245 on Build GRCh38
rs369999291 Chr.5: 112767315-112767315 on Build GRCh38
rs199842850 Chr.5: 112767264-112767264 on Build GRCh38
rs751956779 Chr.5: 112767296-112767296 on Build GRCh38
rs863224279 Chr.5: 112767277-112767277 on Build GRCh38
rs758995578 Chr.5: 112767275-112767275 on Build GRCh38
rs753302494 Chr.5: 112767272-112767272 on Build GRCh38
rs760770847 Chr.5: 112767255-112767255 on Build GRCh38
rs954713888 Chr.5: 112767270-112767270 on Build GRCh38
rs142637152 Chr.5: 112767314-112767314 on Build GRCh38
rs139196838 Chr.5: 112767263-112767263 on Build GRCh38
rs201764637 Chr.5: 112767285-112767285 on Build GRCh38
rs369238363 Chr.5: 112767244-112767244 on Build GRCh38
rs863224542 Chr.5: 112767306-112767306 on Build GRCh38
rs863225335 Chr.5: 112767269-112767269 on Build GRCh38
rs876658977 Chr.5: 112767246-112767246 on Build GRCh38
rs774416950 Chr.5: 112767288-112767288 on Build GRCh38
rs745394881 Chr.5: 112767241-112767241 on Build GRCh38
rs550945533 Chr.5: 112767248-112767248 on Build GRCh38
rs756800793 Chr.5: 112767297-112767297 on Build GRCh38
rs879254143 Chr.5: 112767283-112767286 on Build GRCh38
rs74953290 Chr.5: 112767279-112767279 on Build GRCh38
rs768492450 Chr.5: 112767247-112767247 on Build GRCh38
rs146221748 Chr.5: 112767252-112767252 on Build GRCh38
rs765804855 Chr.5: 112767265-112767265 on Build GRCh38

View Full Product Details