Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25241542-25241704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3A
Entrez Gene ID
1788
Gene Name
DNA methyltransferase 3 alpha
Gene Aliases
DNMT3A2, M.HsaIIIA, TBRS
UniGene
Hs.515840
Gene Chromosome Location
Chr.2: 25232961-25342590 on Build GRCh38
Transcripts
NM_001320893.1 NM_022552.4 NM_153759.3 NM_175629.2 NR_135490.1 XM_005264175.4 XM_005264177.4 XM_011532662.2 XM_011532663.2 XM_011532664.2 XM_011532666.2 XM_011532667.2 XM_017003526.1 XM_017003527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1250494 c.1499G>A p.S500N Chr.2: 25241578-25241578 on Build GRCh38
6907821 c.1411T>G p.Y471D Chr.2: 25241666-25241666 on Build GRCh38
6982406 c.1487G>T p.G496V Chr.2: 25241590-25241590 on Build GRCh38
6907593 c.1502T>A p.V501D Chr.2: 25241575-25241575 on Build GRCh38
6157735 c.2006C>G p.S669C Chr.2: 25241638-25241638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766020170 Chr.2: 25241651-25241651 on Build GRCh38
rs769006337 Chr.2: 25241602-25241602 on Build GRCh38
rs754037033 Chr.2: 25241590-25241590 on Build GRCh38
rs754764419 Chr.2: 25241587-25241587 on Build GRCh38
rs747472028 Chr.2: 25241601-25241601 on Build GRCh38
rs779176507 Chr.2: 25241591-25241591 on Build GRCh38
rs768899154 Chr.2: 25241576-25241576 on Build GRCh38
rs369713081 Chr.2: 25241581-25241581 on Build GRCh38
rs902730598 Chr.2: 25241626-25241626 on Build GRCh38
rs767224028 Chr.2: 25241588-25241588 on Build GRCh38
rs780495518 Chr.2: 25241596-25241596 on Build GRCh38
rs753637583 Chr.2: 25241636-25241637 on Build GRCh38
rs766397390 Chr.2: 25241561-25241561 on Build GRCh38
rs1050271318 Chr.2: 25241612-25241612 on Build GRCh38
rs769539104 Chr.2: 25241617-25241617 on Build GRCh38
rs758490363 Chr.2: 25241594-25241594 on Build GRCh38
rs748613291 Chr.2: 25241615-25241615 on Build GRCh38
rs747168987 Chr.2: 25241685-25241685 on Build GRCh38
rs757328644 Chr.2: 25241639-25241644 on Build GRCh38
rs758402405 Chr.2: 25241678-25241678 on Build GRCh38
rs755960244 Chr.2: 25241578-25241578 on Build GRCh38
rs368961181 Chr.2: 25241675-25241675 on Build GRCh38
rs767552800 Chr.2: 25241665-25241665 on Build GRCh38
rs752434188 Chr.2: 25241668-25241668 on Build GRCh38
rs755982635 Chr.2: 25241669-25241669 on Build GRCh38
rs780120142 Chr.2: 25241680-25241680 on Build GRCh38
rs776843934 Chr.2: 25241610-25241610 on Build GRCh38
rs553080210 Chr.2: 25241656-25241656 on Build GRCh38
rs373549934 Chr.2: 25241586-25241586 on Build GRCh38
rs762503226 Chr.2: 25241650-25241650 on Build GRCh38
rs759408234 Chr.2: 25241660-25241660 on Build GRCh38
rs1025238838 Chr.2: 25241621-25241621 on Build GRCh38
rs146164726 Chr.2: 25241671-25241671 on Build GRCh38
rs763843749 Chr.2: 25241621-25241621 on Build GRCh38
rs375399431 Chr.2: 25241618-25241618 on Build GRCh38

View Full Product Details