Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130862795-130862940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABL1
Entrez Gene ID
25
Gene Name
ABL proto-oncogene 1, non-receptor tyrosine kinase
Gene Aliases
ABL, JTK7, bcr/abl, c-ABL, c-ABL1, p150, v-abl
UniGene
Hs.431048
Gene Chromosome Location
Chr.9: 130713881-130887675 on Build GRCh38
Transcripts
NM_005157.5 NM_007313.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7274261 c.752A>G p.Y251C Chr.9: 130862908-130862908 on Build GRCh38
3779740 c.601C>T p.H201Y Chr.9: 130862814-130862814 on Build GRCh38
4371082 c.749A>G p.N250S Chr.9: 130862905-130862905 on Build GRCh38
93843 c.726C>G p.I242M Chr.9: 130862939-130862939 on Build GRCh38
6591035 c.678C>T p.D226D Chr.9: 130862834-130862834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111371946 Chr.9: 130862903-130862903 on Build GRCh38
rs778982129 Chr.9: 130862885-130862885 on Build GRCh38
rs572239604 Chr.9: 130862905-130862905 on Build GRCh38
rs767096186 Chr.9: 130862831-130862831 on Build GRCh38
rs909306692 Chr.9: 130862848-130862848 on Build GRCh38
rs387906517 Chr.9: 130862919-130862919 on Build GRCh38
rs766252798 Chr.9: 130862849-130862849 on Build GRCh38
rs773957143 Chr.9: 130862819-130862819 on Build GRCh38
rs753573200 Chr.9: 130862871-130862871 on Build GRCh38
rs372729748 Chr.9: 130862834-130862834 on Build GRCh38
rs387906516 Chr.9: 130862920-130862920 on Build GRCh38
rs370646520 Chr.9: 130862824-130862824 on Build GRCh38
rs56271006 Chr.9: 130862872-130862872 on Build GRCh38
rs756057097 Chr.9: 130862843-130862843 on Build GRCh38

View Full Product Details