Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 70965862-70966029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP1
Entrez Gene ID
27086
Gene Name
forkhead box P1
Gene Aliases
12CC4, HSPC215, MFH, QRF1, hFKH1B
UniGene
Hs.59368
Gene Chromosome Location
Chr.3: 70954714-71583989 on Build GRCh38
Transcripts
NM_001244808.1 NM_001244810.1 NM_001244812.1 NM_001244813.1 NM_001244814.1 NM_001244815.1 NM_001244816.1 NM_032682.5 XM_005264735.3 XM_005264736.3 XM_005264737.4 XM_005264742.3 XM_006713102.2 XM_006713103.2 XM_006713104.2 XM_011533584.2 XM_011533585.2 XM_011533588.2 XM_017006165.1 XM_017006166.1 XM_017006167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
23406 c.1808G>A p.R603Q Chr.3: 70965971-70965971 on Build GRCh38
6582728 c.1748_1755delCTCTATAC p.P583fs*34 Chr.3: 70966024-70966031 on Build GRCh38
5355033 c.1889+3_1889+11delGAGTATTGT p.? Chr.3: 70965879-70965887 on Build GRCh38
6098075 c.1812A>G p.E604E Chr.3: 70965967-70965967 on Build GRCh38
5418253 c.1750C>T p.L584L Chr.3: 70966029-70966029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748075031 Chr.3: 70965971-70965971 on Build GRCh38
rs749121207 Chr.3: 70965957-70965957 on Build GRCh38
rs754297044 Chr.3: 70965893-70965893 on Build GRCh38
rs771667463 Chr.3: 70965883-70965883 on Build GRCh38
rs754932182 Chr.3: 70965900-70965900 on Build GRCh38
rs766116383 Chr.3: 70965892-70965892 on Build GRCh38
rs772816590 Chr.3: 70965891-70965891 on Build GRCh38
rs761406403 Chr.3: 70965882-70965882 on Build GRCh38
rs778939994 Chr.3: 70965963-70965963 on Build GRCh38
rs369021347 Chr.3: 70965974-70965974 on Build GRCh38
rs753098583 Chr.3: 70965910-70965910 on Build GRCh38
rs751621278 Chr.3: 70965951-70965951 on Build GRCh38
rs373809702 Chr.3: 70965978-70965978 on Build GRCh38
rs147537388 Chr.3: 70965954-70965954 on Build GRCh38
rs757539300 Chr.3: 70965953-70965953 on Build GRCh38
rs770712857 Chr.3: 70965983-70965983 on Build GRCh38
rs768285337 Chr.3: 70965958-70965958 on Build GRCh38
rs777876809 Chr.3: 70965946-70965946 on Build GRCh38
rs759753513 Chr.3: 70965899-70965899 on Build GRCh38
rs758540752 Chr.3: 70965944-70965944 on Build GRCh38
rs970358469 Chr.3: 70965981-70965981 on Build GRCh38
rs760260048 Chr.3: 70965979-70965979 on Build GRCh38

View Full Product Details