Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99174084-99174241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL11B
Entrez Gene ID
64919
Gene Name
B-cell CLL/lymphoma 11B
Gene Aliases
ATL1, ATL1-alpha, ATL1-beta, ATL1-delta, ATL1-gamma, CTIP-2, CTIP2, RIT1, ZNF856B, hRIT1-alpha
UniGene
Hs.709690
Gene Chromosome Location
Chr.14: 99169287-99271713 on Build GRCh38
Transcripts
NM_001282237.1 NM_001282238.1 NM_022898.2 NM_138576.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1477894 c.2648C>A p.T883N Chr.14: 99174188-99174188 on Build GRCh38
1290347 c.2607G>A p.L869L Chr.14: 99174229-99174229 on Build GRCh38
5745240 c.2624delA p.K875fs*8 Chr.14: 99174212-99174212 on Build GRCh38
6076761 c.2674G>T p.E892* Chr.14: 99174162-99174162 on Build GRCh38
1678246 c.2654A>T p.D885V Chr.14: 99174182-99174182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575421828 Chr.14: 99174149-99174149 on Build GRCh38
rs201198198 Chr.14: 99174214-99174214 on Build GRCh38
rs754460321 Chr.14: 99174107-99174107 on Build GRCh38
rs978705292 Chr.14: 99174136-99174136 on Build GRCh38
rs373415349 Chr.14: 99174145-99174145 on Build GRCh38
rs748104311 Chr.14: 99174142-99174142 on Build GRCh38
rs764796654 Chr.14: 99174190-99174190 on Build GRCh38
rs760505351 Chr.14: 99174147-99174147 on Build GRCh38
rs778094807 Chr.14: 99174121-99174121 on Build GRCh38
rs779336523 Chr.14: 99174110-99174110 on Build GRCh38
rs376796811 Chr.14: 99174211-99174211 on Build GRCh38
rs990764552 Chr.14: 99174205-99174205 on Build GRCh38
rs557237892 Chr.14: 99174135-99174135 on Build GRCh38
rs747313521 Chr.14: 99174125-99174125 on Build GRCh38
rs771778919 Chr.14: 99174137-99174138 on Build GRCh38
rs751946421 Chr.14: 99174219-99174219 on Build GRCh38
rs867667235 Chr.14: 99174217-99174217 on Build GRCh38
rs758752292 Chr.14: 99174120-99174120 on Build GRCh38
rs776324483 Chr.14: 99174184-99174184 on Build GRCh38
rs758918107 Chr.14: 99174204-99174204 on Build GRCh38
rs745988091 Chr.14: 99174134-99174134 on Build GRCh38
rs538615582 Chr.14: 99174133-99174133 on Build GRCh38
rs757505754 Chr.14: 99174131-99174131 on Build GRCh38
rs753213502 Chr.14: 99174117-99174117 on Build GRCh38
rs774221911 Chr.14: 99174137-99174137 on Build GRCh38
rs773146276 Chr.14: 99174146-99174146 on Build GRCh38
rs753198613 Chr.14: 99174196-99174196 on Build GRCh38
rs1040662326 Chr.14: 99174181-99174181 on Build GRCh38
rs759007375 Chr.14: 99174188-99174188 on Build GRCh38

View Full Product Details