Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 29193319-29193484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALK
Entrez Gene ID
238
Gene Name
anaplastic lymphoma receptor tyrosine kinase
Gene Aliases
CD246, NBLST3
UniGene
Hs.654469
Gene Chromosome Location
Chr.2: 29192774-29921611 on Build GRCh38
Transcripts
NM_004304.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6962503 c.1243G>A p.G415S Chr.2: 29193334-29193334 on Build GRCh38
4495941 c.4640C>T p.T1547I Chr.2: 29193447-29193447 on Build GRCh38
7422128 c.4709C>A p.P1570H Chr.2: 29193378-29193378 on Build GRCh38
2941442 c.4627C>T p.P1543S Chr.2: 29193460-29193460 on Build GRCh38
6467182 c.1122C>A p.N374K Chr.2: 29193455-29193455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749162642 Chr.2: 29193353-29193353 on Build GRCh38
rs747946035 Chr.2: 29193344-29193344 on Build GRCh38
rs886055928 Chr.2: 29193439-29193439 on Build GRCh38
rs766508956 Chr.2: 29193338-29193338 on Build GRCh38
rs751872583 Chr.2: 29193407-29193407 on Build GRCh38
rs148351049 Chr.2: 29193364-29193364 on Build GRCh38
rs763403897 Chr.2: 29193455-29193455 on Build GRCh38
rs773870096 Chr.2: 29193383-29193383 on Build GRCh38
rs752927575 Chr.2: 29193401-29193401 on Build GRCh38
rs571671456 Chr.2: 29193398-29193398 on Build GRCh38
rs771622065 Chr.2: 29193460-29193460 on Build GRCh38
rs745390287 Chr.2: 29193430-29193430 on Build GRCh38
rs200829376 Chr.2: 29193358-29193358 on Build GRCh38
rs770217830 Chr.2: 29193367-29193367 on Build GRCh38
rs893771860 Chr.2: 29193362-29193362 on Build GRCh38
rs1021483040 Chr.2: 29193382-29193382 on Build GRCh38
rs767659365 Chr.2: 29193399-29193399 on Build GRCh38
rs751677905 Chr.2: 29193339-29193339 on Build GRCh38
rs199646095 Chr.2: 29193441-29193441 on Build GRCh38
rs78868998 Chr.2: 29193444-29193444 on Build GRCh38
rs878854659 Chr.2: 29193462-29193462 on Build GRCh38
rs35500880 Chr.2: 29193434-29193435 on Build GRCh38
rs745349865 Chr.2: 29193363-29193363 on Build GRCh38
rs774860532 Chr.2: 29193464-29193464 on Build GRCh38
rs763613910 Chr.2: 29193384-29193384 on Build GRCh38
rs952457982 Chr.2: 29193431-29193431 on Build GRCh38
rs751632696 Chr.2: 29193395-29193395 on Build GRCh38
rs771570579 Chr.2: 29193433-29193433 on Build GRCh38
rs773780435 Chr.2: 29193442-29193442 on Build GRCh38
rs200425614 Chr.2: 29193435-29193435 on Build GRCh38
rs542234332 Chr.2: 29193422-29193422 on Build GRCh38
rs150487874 Chr.2: 29193404-29193404 on Build GRCh38
rs1019065457 Chr.2: 29193397-29193397 on Build GRCh38
rs753622072 Chr.2: 29193409-29193409 on Build GRCh38
rs766990201 Chr.2: 29193392-29193392 on Build GRCh38
rs370947313 Chr.2: 29193349-29193349 on Build GRCh38
rs553333897 Chr.2: 29193350-29193350 on Build GRCh38
rs974930400 Chr.2: 29193402-29193402 on Build GRCh38
rs778797863 Chr.2: 29193428-29193428 on Build GRCh38
rs781448587 Chr.2: 29193340-29193340 on Build GRCh38

View Full Product Details