Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69641431-69641555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCND1
Entrez Gene ID
595
Gene Name
cyclin D1
Gene Aliases
BCL1, D11S287E, PRAD1, U21B31
UniGene
Hs.523852
Gene Chromosome Location
Chr.11: 69641105-69654474 on Build GRCh38
Transcripts
NM_053056.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1644296 c.138A>C p.K46N Chr.11: 69641451-69641451 on Build GRCh38
3733241 c.160C>T p.P54S Chr.11: 69641473-69641473 on Build GRCh38
3733356 c.124G>A p.V42M Chr.11: 69641437-69641437 on Build GRCh38
3733343 c.140G>C p.C47S Chr.11: 69641453-69641453 on Build GRCh38
6932624 c.146_148delAGA p.K50delK Chr.11: 69641459-69641461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765904377 Chr.11: 69641479-69641479 on Build GRCh38
rs368364521 Chr.11: 69641524-69641524 on Build GRCh38
rs2930976 Chr.11: 69641469-69641469 on Build GRCh38
rs201441370 Chr.11: 69641530-69641530 on Build GRCh38
rs374405138 Chr.11: 69641515-69641515 on Build GRCh38
rs543211695 Chr.11: 69641527-69641527 on Build GRCh38
rs772785280 Chr.11: 69641466-69641466 on Build GRCh38
rs778678506 Chr.11: 69641530-69641531 on Build GRCh38
rs759345822 Chr.11: 69641510-69641510 on Build GRCh38
rs760341225 Chr.11: 69641474-69641474 on Build GRCh38
rs771611527 Chr.11: 69641457-69641457 on Build GRCh38
rs748632355 Chr.11: 69641498-69641498 on Build GRCh38
rs752676953 Chr.11: 69641516-69641516 on Build GRCh38

View Full Product Details