Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152330653-152330794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW7
Entrez Gene ID
55294
Gene Name
F-box and WD repeat domain containing 7
Gene Aliases
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
UniGene
Hs.561245
Gene Chromosome Location
Chr.4: 152321258-152536095 on Build GRCh38
Transcripts
NM_001013415.1 NM_018315.4 NM_033632.3 XM_011532083.1 XM_011532084.1 XM_011532085.1 XM_011532086.1 XM_011532087.1 XM_011532088.1 XM_017008362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1485742 c.1070G>A p.R357K Chr.4: 152330784-152330784 on Build GRCh38
6191113 c.370_373delACTA p.N125fs*12 Chr.4: 152330764-152330767 on Build GRCh38
6953326 c.566G>A p.W189* Chr.4: 152330760-152330760 on Build GRCh38
4976338 c.373_376delAACT p.N125fs*12 Chr.4: 152330761-152330764 on Build GRCh38
4122774 c.1072C>T p.Q358* Chr.4: 152330782-152330782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568405767 Chr.4: 152330713-152330713 on Build GRCh38
rs112552590 Chr.4: 152330688-152330688 on Build GRCh38
rs367801652 Chr.4: 152330691-152330691 on Build GRCh38
rs754075366 Chr.4: 152330699-152330699 on Build GRCh38
rs761818532 Chr.4: 152330700-152330700 on Build GRCh38
rs750530821 Chr.4: 152330708-152330708 on Build GRCh38
rs745418631 Chr.4: 152330754-152330754 on Build GRCh38
rs777506583 Chr.4: 152330735-152330735 on Build GRCh38
rs775885576 Chr.4: 152330763-152330763 on Build GRCh38
rs761747465 Chr.4: 152330742-152330742 on Build GRCh38
rs748952220 Chr.4: 152330736-152330736 on Build GRCh38
rs755882254 Chr.4: 152330720-152330720 on Build GRCh38
rs202209279 Chr.4: 152330685-152330685 on Build GRCh38
rs937391131 Chr.4: 152330734-152330734 on Build GRCh38
rs760978122 Chr.4: 152330686-152330686 on Build GRCh38
rs560008470 Chr.4: 152330709-152330709 on Build GRCh38
rs772592264 Chr.4: 152330756-152330756 on Build GRCh38
rs999913320 Chr.4: 152330753-152330753 on Build GRCh38
rs201829245 Chr.4: 152330711-152330711 on Build GRCh38

View Full Product Details