Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42256506-42256667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML4
Entrez Gene ID
27436
Gene Name
echinoderm microtubule associated protein like 4
Gene Aliases
C2orf2, ELP120, EMAP-4, EMAPL4, ROPP120
UniGene
Hs.713173
Gene Chromosome Location
Chr.2: 42169338-42332548 on Build GRCh38
Transcripts
NM_001145076.2 NM_019063.4 XM_005264267.1 XM_005264268.3 XM_006711991.1 XM_006711992.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653831 c.224G>A p.R75Q Chr.2: 42256516-42256516 on Build GRCh38
1205263 c.269A>G p.N90S Chr.2: 42256561-42256561 on Build GRCh38
5435362 c.289A>T p.S97C Chr.2: 42256581-42256581 on Build GRCh38
5000366 c.329C>A p.A110D Chr.2: 42256621-42256621 on Build GRCh38
721858 c.223C>T p.R75* Chr.2: 42256515-42256515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151065528 Chr.2: 42256546-42256546 on Build GRCh38
rs763754427 Chr.2: 42256602-42256602 on Build GRCh38
rs766070190 Chr.2: 42256582-42256582 on Build GRCh38
rs762641903 Chr.2: 42256527-42256527 on Build GRCh38
rs534733170 Chr.2: 42256622-42256622 on Build GRCh38
rs367975129 Chr.2: 42256637-42256637 on Build GRCh38
rs749498054 Chr.2: 42256570-42256570 on Build GRCh38
rs373856709 Chr.2: 42256593-42256593 on Build GRCh38
rs149776472 Chr.2: 42256571-42256571 on Build GRCh38
rs772640900 Chr.2: 42256578-42256578 on Build GRCh38
rs773766836 Chr.2: 42256587-42256587 on Build GRCh38
rs779285890 Chr.2: 42256639-42256639 on Build GRCh38
rs549184736 Chr.2: 42256557-42256557 on Build GRCh38
rs761600075 Chr.2: 42256609-42256609 on Build GRCh38
rs372520036 Chr.2: 42256540-42256540 on Build GRCh38
rs774899854 Chr.2: 42256594-42256594 on Build GRCh38
rs767451716 Chr.2: 42256550-42256550 on Build GRCh38
rs751385923 Chr.2: 42256544-42256544 on Build GRCh38
rs750359091 Chr.2: 42256632-42256632 on Build GRCh38
rs762549058 Chr.2: 42256599-42256599 on Build GRCh38
rs142252734 Chr.2: 42256573-42256573 on Build GRCh38
rs753887338 Chr.2: 42256636-42256636 on Build GRCh38
rs748537313 Chr.2: 42256576-42256576 on Build GRCh38
rs750400296 Chr.2: 42256562-42256562 on Build GRCh38
rs774073765 Chr.2: 42256603-42256603 on Build GRCh38
rs756048433 Chr.2: 42256635-42256635 on Build GRCh38
rs780048815 Chr.2: 42256569-42256569 on Build GRCh38
rs747530990 Chr.2: 42256589-42256589 on Build GRCh38

View Full Product Details