Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207555564-207555712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB1
Entrez Gene ID
1385
Gene Name
cAMP responsive element binding protein 1
Gene Aliases
CREB, CREB-1
UniGene
Hs.516646
Gene Chromosome Location
Chr.2: 207529892-207605989 on Build GRCh38
Transcripts
NM_001320793.1 NM_004379.4 NM_134442.4 NR_135473.1 XM_011510645.1 XM_011510646.2 XM_011510647.2 XM_011510648.2 XM_011510649.2 XM_017003399.1 XM_017003400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2714427 c.45A>T p.A15A Chr.2: 207555680-207555680 on Build GRCh38
2714426 c.24G>C p.E8D Chr.2: 207555659-207555659 on Build GRCh38
6198632 c.3G>A p.M1I Chr.2: 207555638-207555638 on Build GRCh38
2714428 c.66C>T p.N22N Chr.2: 207555701-207555701 on Build GRCh38
4881775 c.37G>A p.G13R Chr.2: 207555672-207555672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936563196 Chr.2: 207555683-207555684 on Build GRCh38
rs766571257 Chr.2: 207555676-207555676 on Build GRCh38
rs1035168298 Chr.2: 207555603-207555603 on Build GRCh38
rs772288583 Chr.2: 207555629-207555629 on Build GRCh38
rs769252949 Chr.2: 207555593-207555593 on Build GRCh38
rs775337117 Chr.2: 207555682-207555682 on Build GRCh38
rs35318842 Chr.2: 207555643-207555644 on Build GRCh38
rs961180040 Chr.2: 207555616-207555616 on Build GRCh38
rs991071029 Chr.2: 207555670-207555670 on Build GRCh38
rs367727829 Chr.2: 207555657-207555657 on Build GRCh38
rs775027151 Chr.2: 207555601-207555601 on Build GRCh38
rs544455173 Chr.2: 207555592-207555592 on Build GRCh38
rs773236696 Chr.2: 207555630-207555630 on Build GRCh38
rs748187730 Chr.2: 207555609-207555609 on Build GRCh38
rs771400764 Chr.2: 207555614-207555614 on Build GRCh38
rs751134810 Chr.2: 207555593-207555594 on Build GRCh38

View Full Product Details