Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150054008-150054150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF1R
Entrez Gene ID
1436
Gene Name
colony stimulating factor 1 receptor
Gene Aliases
C-FMS, CD115, CSF-1R, CSFR, FIM2, FMS, HDLS, M-CSF-R
UniGene
Hs.586219
Gene Chromosome Location
Chr.5: 150053291-150113372 on Build GRCh38
Transcripts
NM_001288705.1 NM_005211.3 NR_109969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
950 c.2905T>G p.Y969D Chr.5: 150054083-150054083 on Build GRCh38
6636025 c.2879C>T p.A960V Chr.5: 150054109-150054109 on Build GRCh38
6924794 c.2845G>A p.E949K Chr.5: 150054143-150054143 on Build GRCh38
951 c.2905T>A p.Y969N Chr.5: 150054083-150054083 on Build GRCh38
952 c.2905T>C p.Y969H Chr.5: 150054083-150054083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2066933 Chr.5: 150054034-150054034 on Build GRCh38
rs756779459 Chr.5: 150054030-150054030 on Build GRCh38
rs376038713 Chr.5: 150054065-150054065 on Build GRCh38
rs547653185 Chr.5: 150054060-150054060 on Build GRCh38
rs761591053 Chr.5: 150054113-150054113 on Build GRCh38
rs370648666 Chr.5: 150054102-150054102 on Build GRCh38
rs121913393 Chr.5: 150054083-150054083 on Build GRCh38
rs766047383 Chr.5: 150054078-150054079 on Build GRCh38
rs766007425 Chr.5: 150054128-150054128 on Build GRCh38
rs764837673 Chr.5: 150054115-150054115 on Build GRCh38
rs2066934 Chr.5: 150054033-150054033 on Build GRCh38
rs1801271 Chr.5: 150054082-150054082 on Build GRCh38
rs768534817 Chr.5: 150054107-150054107 on Build GRCh38
rs121913392 Chr.5: 150054081-150054081 on Build GRCh38
rs751974904 Chr.5: 150054130-150054130 on Build GRCh38
rs200068110 Chr.5: 150054125-150054125 on Build GRCh38
rs56005231 Chr.5: 150054126-150054126 on Build GRCh38
rs35308019 Chr.5: 150054033-150054033 on Build GRCh38
rs749542020 Chr.5: 150054068-150054068 on Build GRCh38
rs370469459 Chr.5: 150054045-150054045 on Build GRCh38
rs775465143 Chr.5: 150054085-150054085 on Build GRCh38
rs894716510 Chr.5: 150054064-150054064 on Build GRCh38
rs771254626 Chr.5: 150054072-150054072 on Build GRCh38
rs776645465 Chr.5: 150054109-150054109 on Build GRCh38
rs764596420 Chr.5: 150054031-150054031 on Build GRCh38
rs386693509 Chr.5: 150054033-150054034 on Build GRCh38
rs374082775 Chr.5: 150054059-150054059 on Build GRCh38
rs879603703 Chr.5: 150054033-150054034 on Build GRCh38

View Full Product Details