Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130862661-130862833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABL1
Entrez Gene ID
25
Gene Name
ABL proto-oncogene 1, non-receptor tyrosine kinase
Gene Aliases
ABL, JTK7, bcr/abl, c-ABL, c-ABL1, p150, v-abl
UniGene
Hs.431048
Gene Chromosome Location
Chr.9: 130713881-130887675 on Build GRCh38
Transcripts
NM_005157.5 NM_007313.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6945255 c.638C>T p.T213I Chr.9: 130862794-130862794 on Build GRCh38
3779740 c.601C>T p.H201Y Chr.9: 130862814-130862814 on Build GRCh38
6961509 c.608T>G p.L203R Chr.9: 130862764-130862764 on Build GRCh38
3654897 c.617C>T p.A206V Chr.9: 130862830-130862830 on Build GRCh38
6182777 c.612G>C p.T204T Chr.9: 130862825-130862825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368646202 Chr.9: 130862784-130862784 on Build GRCh38
rs1044402373 Chr.9: 130862688-130862688 on Build GRCh38
rs767535809 Chr.9: 130862748-130862748 on Build GRCh38
rs766685037 Chr.9: 130862718-130862718 on Build GRCh38
rs534700488 Chr.9: 130862693-130862693 on Build GRCh38
rs775242844 Chr.9: 130862734-130862734 on Build GRCh38
rs1031931287 Chr.9: 130862704-130862704 on Build GRCh38
rs7856784 Chr.9: 130862687-130862687 on Build GRCh38
rs755144370 Chr.9: 130862785-130862785 on Build GRCh38
rs556586112 Chr.9: 130862791-130862791 on Build GRCh38
rs762449425 Chr.9: 130862736-130862736 on Build GRCh38
rs1017846667 Chr.9: 130862752-130862752 on Build GRCh38
rs146008880 Chr.9: 130862801-130862801 on Build GRCh38
rs964586142 Chr.9: 130862754-130862754 on Build GRCh38
rs756003964 Chr.9: 130862769-130862769 on Build GRCh38
rs375905680 Chr.9: 130862739-130862739 on Build GRCh38
rs761005394 Chr.9: 130862715-130862715 on Build GRCh38
rs976094954 Chr.9: 130862810-130862810 on Build GRCh38
rs150134901 Chr.9: 130862802-130862802 on Build GRCh38
rs79052735 Chr.9: 130862772-130862772 on Build GRCh38
rs763676375 Chr.9: 130862737-130862737 on Build GRCh38
rs1027274279 Chr.9: 130862768-130862768 on Build GRCh38
rs772617122 Chr.9: 130862799-130862799 on Build GRCh38
rs779117534 Chr.9: 130862790-130862790 on Build GRCh38
rs779645963 Chr.9: 130862774-130862774 on Build GRCh38
rs537025188 Chr.9: 130862742-130862742 on Build GRCh38
rs750336311 Chr.9: 130862756-130862756 on Build GRCh38
rs538889953 Chr.9: 130862805-130862805 on Build GRCh38
rs776806158 Chr.9: 130862803-130862803 on Build GRCh38
rs869199784 Chr.9: 130862753-130862753 on Build GRCh38
rs567943403 Chr.9: 130862703-130862703 on Build GRCh38
rs548204394 Chr.9: 130862702-130862702 on Build GRCh38

View Full Product Details