Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 151880536-151880706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESR1
Entrez Gene ID
2099
Gene Name
estrogen receptor 1
Gene Aliases
ER, ESR, ESRA, ESTRR, Era, NR3A1
UniGene
Hs.208124
Gene Chromosome Location
Chr.6: 151654148-152103274 on Build GRCh38
Transcripts
NM_000125.3 NM_001122740.1 NM_001122741.1 NM_001122742.1 NM_001291230.1 NM_001291241.1 NM_001328100.1 XM_006715374.3 XM_006715375.3 XM_011535543.2 XM_011535544.2 XM_011535545.2 XM_011535547.2 XM_017010376.1 XM_017010377.1 XM_017010378.1 XM_017010379.1 XM_017010380.1 XM_017010381.1 XM_017010382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6934011 c.152_153CC>TT p.T51I Chr.6: 151880682-151880683 on Build GRCh38
6918924 c.682A>G p.T228A Chr.6: 151880693-151880693 on Build GRCh38
6918925 c.163A>G p.T55A Chr.6: 151880693-151880693 on Build GRCh38
3430123 c.650A>G p.N217S Chr.6: 151880661-151880661 on Build GRCh38
6984446 c.133G>A p.D45N Chr.6: 151880663-151880663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027310877 Chr.6: 151880635-151880635 on Build GRCh38
rs769383399 Chr.6: 151880639-151880639 on Build GRCh38
rs761613029 Chr.6: 151880663-151880663 on Build GRCh38
rs762803413 Chr.6: 151880610-151880611 on Build GRCh38
rs748537766 Chr.6: 151880624-151880624 on Build GRCh38
rs768352205 Chr.6: 151880658-151880658 on Build GRCh38
rs762739348 Chr.6: 151880656-151880656 on Build GRCh38
rs878864568 Chr.6: 151880563-151880563 on Build GRCh38
rs556769274 Chr.6: 151880591-151880591 on Build GRCh38
rs778281774 Chr.6: 151880628-151880628 on Build GRCh38
rs545480471 Chr.6: 151880669-151880669 on Build GRCh38
rs370499721 Chr.6: 151880627-151880627 on Build GRCh38
rs201978526 Chr.6: 151880637-151880637 on Build GRCh38
rs75069603 Chr.6: 151880662-151880662 on Build GRCh38
rs775122947 Chr.6: 151880649-151880649 on Build GRCh38
rs745405487 Chr.6: 151880634-151880634 on Build GRCh38
rs535167918 Chr.6: 151880582-151880582 on Build GRCh38

View Full Product Details