Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 395870-396043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF4
Entrez Gene ID
3662
Gene Name
interferon regulatory factor 4
Gene Aliases
LSIRF, MUM1, NF-EM5, SHEP8
UniGene
Hs.401013
Gene Chromosome Location
Chr.6: 391739-411443 on Build GRCh38
Transcripts
NM_001195286.1 NM_002460.3 NR_046000.2 XM_006715090.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6977092 c.427G>A p.D143N Chr.6: 395870-395870 on Build GRCh38
6964138 c.451C>T p.P151S Chr.6: 395894-395894 on Build GRCh38
6958732 c.484C>T p.P162S Chr.6: 395927-395927 on Build GRCh38
7224078 c.476C>G p.P159R Chr.6: 395919-395919 on Build GRCh38
6967559 c.452C>T p.P151L Chr.6: 395895-395895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781044692 Chr.6: 395959-395959 on Build GRCh38
rs148022406 Chr.6: 395923-395923 on Build GRCh38
rs1022665553 Chr.6: 396011-396011 on Build GRCh38
rs374802152 Chr.6: 395958-395958 on Build GRCh38
rs771434117 Chr.6: 395920-395920 on Build GRCh38
rs755460615 Chr.6: 395910-395910 on Build GRCh38
rs761027628 Chr.6: 395902-395902 on Build GRCh38
rs990234547 Chr.6: 396008-396008 on Build GRCh38
rs73717071 Chr.6: 395889-395889 on Build GRCh38
rs766413794 Chr.6: 395927-395927 on Build GRCh38
rs751600532 Chr.6: 395929-395929 on Build GRCh38
rs750505973 Chr.6: 395916-395916 on Build GRCh38
rs199665380 Chr.6: 395967-395967 on Build GRCh38
rs978687745 Chr.6: 395993-395993 on Build GRCh38
rs1032009237 Chr.6: 395978-395978 on Build GRCh38
rs765193356 Chr.6: 395911-395911 on Build GRCh38
rs764577689 Chr.6: 395903-395903 on Build GRCh38
rs748767237 Chr.6: 395987-395987 on Build GRCh38
rs959712797 Chr.6: 395940-395940 on Build GRCh38
rs748167619 Chr.6: 395992-395992 on Build GRCh38
rs144432479 Chr.6: 395996-395996 on Build GRCh38
rs777342323 Chr.6: 395984-395984 on Build GRCh38
rs776019676 Chr.6: 395890-395890 on Build GRCh38
rs755812276 Chr.6: 395969-395969 on Build GRCh38
rs201185528 Chr.6: 395891-395891 on Build GRCh38
rs934455833 Chr.6: 396020-396020 on Build GRCh38
rs776661032 Chr.6: 395906-395906 on Build GRCh38

View Full Product Details