Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152322976-152323127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW7
Entrez Gene ID
55294
Gene Name
F-box and WD repeat domain containing 7
Gene Aliases
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
UniGene
Hs.561245
Gene Chromosome Location
Chr.4: 152321258-152536095 on Build GRCh38
Transcripts
NM_001013415.1 NM_018315.4 NM_033632.3 XM_011532083.1 XM_011532084.1 XM_011532085.1 XM_011532086.1 XM_011532087.1 XM_011532088.1 XM_017008362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5344633 c.1931G>A p.G644E Chr.4: 152323074-152323074 on Build GRCh38
480953 c.1782G>C p.R594R Chr.4: 152322983-152322983 on Build GRCh38
6662504 c.1739delT p.L580fs*47 Chr.4: 152323026-152323026 on Build GRCh38
7262887 c.1215A>G p.G405G Chr.4: 152323073-152323073 on Build GRCh38
4837614 c.1618C>A p.R540R Chr.4: 152323033-152323033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7679116 Chr.4: 152323003-152323003 on Build GRCh38
rs749701779 Chr.4: 152323004-152323004 on Build GRCh38
rs200198533 Chr.4: 152323082-152323082 on Build GRCh38
rs957874517 Chr.4: 152323011-152323011 on Build GRCh38
rs201270631 Chr.4: 152323019-152323019 on Build GRCh38
rs765315069 Chr.4: 152323094-152323094 on Build GRCh38
rs961642075 Chr.4: 152323002-152323002 on Build GRCh38
rs761828621 Chr.4: 152323061-152323061 on Build GRCh38
rs144247898 Chr.4: 152323033-152323033 on Build GRCh38
rs34709778 Chr.4: 152323075-152323076 on Build GRCh38
rs1029868423 Chr.4: 152323072-152323072 on Build GRCh38
rs767500139 Chr.4: 152323046-152323046 on Build GRCh38
rs759610249 Chr.4: 152323032-152323032 on Build GRCh38
rs775781675 Chr.4: 152323047-152323047 on Build GRCh38
rs998428097 Chr.4: 152323053-152323053 on Build GRCh38
rs974902950 Chr.4: 152323021-152323021 on Build GRCh38
rs750215508 Chr.4: 152323099-152323099 on Build GRCh38

View Full Product Details