Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 151842703-151842846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESR1
Entrez Gene ID
2099
Gene Name
estrogen receptor 1
Gene Aliases
ER, ESR, ESRA, ESTRR, Era, NR3A1
UniGene
Hs.208124
Gene Chromosome Location
Chr.6: 151654148-152103274 on Build GRCh38
Transcripts
NM_000125.3 NM_001122740.1 NM_001122741.1 NM_001122742.1 NM_001291230.1 NM_001291241.1 NM_001328100.1 XM_006715374.3 XM_006715375.3 XM_011535543.2 XM_011535544.2 XM_011535545.2 XM_011535547.2 XM_017010376.1 XM_017010377.1 XM_017010378.1 XM_017010379.1 XM_017010380.1 XM_017010381.1 XM_017010382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4428865 c.78C>T p.V26V Chr.6: 151842741-151842741 on Build GRCh38
4503502 c.121C>T p.Q41* Chr.6: 151842784-151842784 on Build GRCh38
4992439 c.630G>A p.K210K Chr.6: 151842774-151842774 on Build GRCh38
6906869 c.640C>G p.Q214E Chr.6: 151842784-151842784 on Build GRCh38
1205680 c.632G>T p.R211I Chr.6: 151842776-151842776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762866918 Chr.6: 151842794-151842794 on Build GRCh38
rs776069453 Chr.6: 151842739-151842739 on Build GRCh38
rs969753075 Chr.6: 151842792-151842792 on Build GRCh38
rs564461683 Chr.6: 151842793-151842793 on Build GRCh38
rs376774696 Chr.6: 151842814-151842814 on Build GRCh38
rs1023093115 Chr.6: 151842747-151842747 on Build GRCh38
rs763847841 Chr.6: 151842802-151842802 on Build GRCh38
rs533357620 Chr.6: 151842806-151842806 on Build GRCh38
rs556478419 Chr.6: 151842732-151842732 on Build GRCh38
rs759101568 Chr.6: 151842782-151842782 on Build GRCh38
rs767478932 Chr.6: 151842811-151842811 on Build GRCh38
rs757209337 Chr.6: 151842807-151842807 on Build GRCh38
rs750516960 Chr.6: 151842812-151842812 on Build GRCh38

View Full Product Details