Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96191929-96192078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLNK
Entrez Gene ID
29760
Gene Name
B-cell linker
Gene Aliases
AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca
UniGene
Hs.665244
Gene Chromosome Location
Chr.10: 96189171-96271576 on Build GRCh38
Transcripts
NM_001114094.1 NM_001258440.1 NM_001258441.1 NM_001258442.1 NM_013314.3 NR_047680.1 NR_047681.1 NR_047682.1 NR_047683.1 XM_011539728.2 XM_017016159.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
428210 c.1275C>T p.I425I Chr.10: 96192069-96192069 on Build GRCh38
4815618 c.1170C>T p.I390I Chr.10: 96192069-96192069 on Build GRCh38
921885 c.1283A>C p.N428T Chr.10: 96192061-96192061 on Build GRCh38
1246134 c.1362A>C p.K454N Chr.10: 96191982-96191982 on Build GRCh38
4870634 c.1228G>T p.D410Y Chr.10: 96192011-96192011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782445849 Chr.10: 96192028-96192028 on Build GRCh38
rs200396708 Chr.10: 96191966-96191966 on Build GRCh38
rs781847413 Chr.10: 96191971-96191971 on Build GRCh38
rs782736210 Chr.10: 96191969-96191969 on Build GRCh38
rs931320409 Chr.10: 96192018-96192018 on Build GRCh38
rs781802342 Chr.10: 96192024-96192024 on Build GRCh38
rs782636828 Chr.10: 96192042-96192042 on Build GRCh38
rs782643739 Chr.10: 96191958-96191959 on Build GRCh38
rs782515286 Chr.10: 96191959-96191959 on Build GRCh38
rs1057940 Chr.10: 96192005-96192005 on Build GRCh38
rs949259126 Chr.10: 96191965-96191965 on Build GRCh38
rs782280453 Chr.10: 96192025-96192025 on Build GRCh38
rs782660063 Chr.10: 96192010-96192010 on Build GRCh38
rs782608058 Chr.10: 96192033-96192033 on Build GRCh38
rs782547336 Chr.10: 96191993-96191993 on Build GRCh38
rs762002095 Chr.10: 96192030-96192030 on Build GRCh38
rs918547258 Chr.10: 96191967-96191967 on Build GRCh38
rs781794307 Chr.10: 96191964-96191964 on Build GRCh38
rs782710142 Chr.10: 96191950-96191950 on Build GRCh38

View Full Product Details