Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 151808083-151808252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESR1
Entrez Gene ID
2099
Gene Name
estrogen receptor 1
Gene Aliases
ER, ESR, ESRA, ESTRR, Era, NR3A1
UniGene
Hs.208124
Gene Chromosome Location
Chr.6: 151654148-152103274 on Build GRCh38
Transcripts
NM_000125.3 NM_001122740.1 NM_001122741.1 NM_001122742.1 NM_001291230.1 NM_001291241.1 XM_006715374.3 XM_011535543.2 XM_011535544.2 XM_011535545.2 XM_011535547.2 XM_017010376.1 XM_017010377.1 XM_017010378.1 XM_017010379.1 XM_017010380.1 XM_017010381.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1441225 c.202G>A p.A68T Chr.6: 151808114-151808114 on Build GRCh38
5345063 c.220G>A p.G74S Chr.6: 151808132-151808132 on Build GRCh38
5641375 c.339G>C p.P113P Chr.6: 151808251-151808251 on Build GRCh38
1441226 c.240delC p.Y80fs*1 Chr.6: 151808152-151808152 on Build GRCh38
5656533 c.267C>A p.F89L Chr.6: 151808179-151808179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767863538 Chr.6: 151808207-151808207 on Build GRCh38
rs773500294 Chr.6: 151808208-151808208 on Build GRCh38
rs956938848 Chr.6: 151808174-151808174 on Build GRCh38
rs1018021627 Chr.6: 151808107-151808107 on Build GRCh38
rs749296901 Chr.6: 151808200-151808200 on Build GRCh38
rs746521050 Chr.6: 151808181-151808181 on Build GRCh38
rs771540162 Chr.6: 151808114-151808114 on Build GRCh38
rs746432 Chr.6: 151808173-151808173 on Build GRCh38
rs780595074 Chr.6: 151808100-151808100 on Build GRCh38
rs745670953 Chr.6: 151808193-151808193 on Build GRCh38
rs767847192 Chr.6: 151808144-151808144 on Build GRCh38
rs756964962 Chr.6: 151808189-151808189 on Build GRCh38
rs747623367 Chr.6: 151808111-151808111 on Build GRCh38
rs775435150 Chr.6: 151808132-151808132 on Build GRCh38
rs777319857 Chr.6: 151808116-151808116 on Build GRCh38
rs766714686 Chr.6: 151808219-151808219 on Build GRCh38
rs964351047 Chr.6: 151808193-151808194 on Build GRCh38
rs750873681 Chr.6: 151808150-151808150 on Build GRCh38
rs754327679 Chr.6: 151808158-151808158 on Build GRCh38
rs905397493 Chr.6: 151808135-151808135 on Build GRCh38
rs533291788 Chr.6: 151808203-151808203 on Build GRCh38
rs776480169 Chr.6: 151808121-151808121 on Build GRCh38
rs181468071 Chr.6: 151808227-151808227 on Build GRCh38
rs746640340 Chr.6: 151808139-151808139 on Build GRCh38
rs941006547 Chr.6: 151808176-151808176 on Build GRCh38
rs975546721 Chr.6: 151808226-151808226 on Build GRCh38
rs146586199 Chr.6: 151808119-151808119 on Build GRCh38
rs761971425 Chr.6: 151808206-151808206 on Build GRCh38
rs572261788 Chr.6: 151808151-151808151 on Build GRCh38
rs762165734 Chr.6: 151808143-151808143 on Build GRCh38
rs1019644335 Chr.6: 151808209-151808209 on Build GRCh38
rs775313423 Chr.6: 151808199-151808199 on Build GRCh38
rs769593068 Chr.6: 151808196-151808196 on Build GRCh38
rs759317032 Chr.6: 151808125-151808125 on Build GRCh38
rs756487587 Chr.6: 151808153-151808153 on Build GRCh38
rs754125870 Chr.6: 151808225-151808225 on Build GRCh38
rs769634976 Chr.6: 151808130-151808130 on Build GRCh38
rs561015454 Chr.6: 151808228-151808228 on Build GRCh38
rs9340773 Chr.6: 151808141-151808141 on Build GRCh38
rs766797889 Chr.6: 151808154-151808154 on Build GRCh38

View Full Product Details