Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78216603-78216738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC5
Entrez Gene ID
332
Gene Name
baculoviral IAP repeat containing 5
Gene Aliases
API4, EPR-1
UniGene
Hs.744872
Gene Chromosome Location
Chr.17: 78214196-78225635 on Build GRCh38
Transcripts
NM_001012270.1 NM_001012271.1 NM_001168.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4831772 c.851C>G p.S284C Chr.17: 78216665-78216665 on Build GRCh38
7156033 c.349G>A p.E117K Chr.17: 78216722-78216722 on Build GRCh38
7205934 c.254_255insGGTTGCG p.F86fs*11 Chr.17: 78216696-78216697 on Build GRCh38
7205933 c.323_324insGGTTGCG p.F109fs*11 Chr.17: 78216696-78216697 on Build GRCh38
3522790 c.312G>C p.S104S Chr.17: 78216685-78216685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751306383 Chr.17: 78216637-78216637 on Build GRCh38
rs772358465 Chr.17: 78216645-78216645 on Build GRCh38
rs750736960 Chr.17: 78216684-78216684 on Build GRCh38
rs762005531 Chr.17: 78216671-78216671 on Build GRCh38
rs755408216 Chr.17: 78216694-78216694 on Build GRCh38
rs183808064 Chr.17: 78216643-78216643 on Build GRCh38
rs769299439 Chr.17: 78216641-78216641 on Build GRCh38
rs747598585 Chr.17: 78216695-78216695 on Build GRCh38
rs376560568 Chr.17: 78216707-78216707 on Build GRCh38
rs371741114 Chr.17: 78216642-78216642 on Build GRCh38
rs761040974 Chr.17: 78216652-78216652 on Build GRCh38
rs775794750 Chr.17: 78216710-78216710 on Build GRCh38
rs765360796 Chr.17: 78216679-78216679 on Build GRCh38
rs200189113 Chr.17: 78216689-78216689 on Build GRCh38
rs371975672 Chr.17: 78216685-78216685 on Build GRCh38
rs754830798 Chr.17: 78216690-78216690 on Build GRCh38
rs368114063 Chr.17: 78216665-78216665 on Build GRCh38
rs764695425 Chr.17: 78216655-78216655 on Build GRCh38
rs775847864 Chr.17: 78216648-78216648 on Build GRCh38
rs777270514 Chr.17: 78216697-78216697 on Build GRCh38
rs748883772 Chr.17: 78216703-78216703 on Build GRCh38
rs766217359 Chr.17: 78216688-78216688 on Build GRCh38

View Full Product Details