Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41578052-41578198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP4
Entrez Gene ID
116113
Gene Name
forkhead box P4
Gene Aliases
hFKHLA
UniGene
Hs.131436
Gene Chromosome Location
Chr.6: 41546426-41602384 on Build GRCh38
Transcripts
NM_001012426.1 NM_001012427.1 NM_138457.2 XM_006714991.3 XM_011514289.2 XM_011514290.2 XM_011514291.2 XM_011514292.2 XM_017010233.1 XM_017010234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
741932 c.281A>T p.Q94L Chr.6: 41578062-41578062 on Build GRCh38
7177597 c.271G>A p.D91N Chr.6: 41578052-41578052 on Build GRCh38
741933 c.281A>T p.Q94L Chr.6: 41578062-41578062 on Build GRCh38
7279579 c.293C>T p.A98V Chr.6: 41578074-41578074 on Build GRCh38
7279578 c.293C>T p.A98V Chr.6: 41578074-41578074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907317362 Chr.6: 41578156-41578156 on Build GRCh38
rs138103012 Chr.6: 41578101-41578101 on Build GRCh38
rs554628504 Chr.6: 41578157-41578157 on Build GRCh38
rs559444252 Chr.6: 41578107-41578107 on Build GRCh38
rs998132867 Chr.6: 41578123-41578123 on Build GRCh38
rs956369643 Chr.6: 41578162-41578162 on Build GRCh38
rs534672425 Chr.6: 41578153-41578153 on Build GRCh38
rs772077879 Chr.6: 41578171-41578171 on Build GRCh38
rs376541506 Chr.6: 41578085-41578085 on Build GRCh38
rs1046642754 Chr.6: 41578108-41578108 on Build GRCh38
rs756620453 Chr.6: 41578125-41578125 on Build GRCh38
rs372517776 Chr.6: 41578075-41578075 on Build GRCh38
rs765880796 Chr.6: 41578097-41578097 on Build GRCh38
rs760590284 Chr.6: 41578094-41578094 on Build GRCh38
rs749636290 Chr.6: 41578097-41578097 on Build GRCh38
rs746393065 Chr.6: 41578124-41578124 on Build GRCh38
rs200593493 Chr.6: 41578105-41578105 on Build GRCh38
rs370592290 Chr.6: 41578112-41578112 on Build GRCh38

View Full Product Details