Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85648525-85648792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSE1
Entrez Gene ID
23199
Gene Name
Gse1 coiled-coil protein
Gene Aliases
CRHSP24, KIAA0182
UniGene
Hs.461647
Gene Chromosome Location
Chr.16: 85169512-85676206 on Build GRCh38
Transcripts
NM_001134473.2 NM_001278184.1 NM_014615.3 XM_005255859.4 XM_005255860.3 XM_005255861.4 XM_005255863.4 XM_005255864.4 XM_005255865.4 XM_011522965.2 XM_011522966.2 XM_017023083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4695140 c.304C>T p.R102C Chr.16: 85648629-85648629 on Build GRCh38
4695139 c.288C>T p.P96P Chr.16: 85648613-85648613 on Build GRCh38
3513068 c.378C>T p.I126I Chr.16: 85648703-85648703 on Build GRCh38
6450965 c.328G>A p.V110I Chr.16: 85648653-85648653 on Build GRCh38
6044810 c.359_360insCC p.V123fs*3 Chr.16: 85648684-85648685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781202749 Chr.16: 85648742-85648742 on Build GRCh38
rs532644893 Chr.16: 85648703-85648703 on Build GRCh38
rs768214662 Chr.16: 85648629-85648629 on Build GRCh38
rs570962793 Chr.16: 85648552-85648552 on Build GRCh38
rs762078512 Chr.16: 85648735-85648735 on Build GRCh38
rs766630200 Chr.16: 85648632-85648632 on Build GRCh38
rs770603470 Chr.16: 85648610-85648610 on Build GRCh38
rs369354583 Chr.16: 85648646-85648646 on Build GRCh38
rs745680419 Chr.16: 85648673-85648673 on Build GRCh38
rs1051200506 Chr.16: 85648575-85648575 on Build GRCh38
rs150142654 Chr.16: 85648598-85648598 on Build GRCh38
rs969150216 Chr.16: 85648630-85648630 on Build GRCh38
rs368435782 Chr.16: 85648652-85648652 on Build GRCh38
rs756423088 Chr.16: 85648586-85648586 on Build GRCh38
rs137897923 Chr.16: 85648729-85648729 on Build GRCh38
rs539912933 Chr.16: 85648692-85648692 on Build GRCh38
rs768318723 Chr.16: 85648747-85648747 on Build GRCh38
rs149283578 Chr.16: 85648659-85648659 on Build GRCh38
rs961969619 Chr.16: 85648543-85648543 on Build GRCh38
rs770263163 Chr.16: 85648763-85648763 on Build GRCh38
rs779863020 Chr.16: 85648746-85648746 on Build GRCh38
rs143043046 Chr.16: 85648748-85648748 on Build GRCh38
rs1052711033 Chr.16: 85648766-85648766 on Build GRCh38
rs780813332 Chr.16: 85648669-85648669 on Build GRCh38
rs972296006 Chr.16: 85648558-85648558 on Build GRCh38
rs143440811 Chr.16: 85648660-85648660 on Build GRCh38
rs773702179 Chr.16: 85648631-85648631 on Build GRCh38
rs1023386528 Chr.16: 85648624-85648624 on Build GRCh38
rs750386270 Chr.16: 85648744-85648744 on Build GRCh38
rs757080641 Chr.16: 85648736-85648736 on Build GRCh38
rs369786356 Chr.16: 85648762-85648762 on Build GRCh38
rs923152991 Chr.16: 85648565-85648565 on Build GRCh38
rs776541218 Chr.16: 85648611-85648611 on Build GRCh38
rs911305616 Chr.16: 85648724-85648724 on Build GRCh38
rs997151009 Chr.16: 85648717-85648717 on Build GRCh38
rs533050963 Chr.16: 85648691-85648691 on Build GRCh38
rs771280191 Chr.16: 85648757-85648757 on Build GRCh38
rs865981619 Chr.16: 85648670-85648670 on Build GRCh38
rs761037151 Chr.16: 85648690-85648690 on Build GRCh38
rs138417000 Chr.16: 85648653-85648653 on Build GRCh38
rs371776973 Chr.16: 85648614-85648614 on Build GRCh38
rs779669057 Chr.16: 85648676-85648676 on Build GRCh38
rs949717861 Chr.16: 85648655-85648655 on Build GRCh38
rs759633198 Chr.16: 85648643-85648643 on Build GRCh38
rs373579949 Chr.16: 85648581-85648581 on Build GRCh38
rs755276325 Chr.16: 85648594-85648594 on Build GRCh38
rs35869664 Chr.16: 85648568-85648568 on Build GRCh38
rs746998295 Chr.16: 85648609-85648609 on Build GRCh38
rs564482434 Chr.16: 85648688-85648688 on Build GRCh38
rs754199513 Chr.16: 85648684-85648684 on Build GRCh38
rs201252847 Chr.16: 85648760-85648760 on Build GRCh38
rs370456740 Chr.16: 85648574-85648574 on Build GRCh38
rs761640820 Chr.16: 85648546-85648546 on Build GRCh38
rs1054280312 Chr.16: 85648685-85648685 on Build GRCh38
rs75392774 Chr.16: 85648592-85648592 on Build GRCh38
rs769272935 Chr.16: 85648674-85648674 on Build GRCh38
rs374934041 Chr.16: 85648635-85648635 on Build GRCh38
rs754251796 Chr.16: 85648636-85648636 on Build GRCh38
rs375370402 Chr.16: 85648615-85648615 on Build GRCh38
rs35744900 Chr.16: 85648661-85648662 on Build GRCh38
rs553403743 Chr.16: 85648580-85648580 on Build GRCh38
rs764524595 Chr.16: 85648683-85648684 on Build GRCh38
rs201812312 Chr.16: 85648613-85648613 on Build GRCh38
rs777863962 Chr.16: 85648608-85648608 on Build GRCh38
rs896610470 Chr.16: 85648767-85648767 on Build GRCh38
rs371934256 Chr.16: 85648672-85648672 on Build GRCh38
rs376778699 Chr.16: 85648745-85648745 on Build GRCh38
rs1013720410 Chr.16: 85648619-85648619 on Build GRCh38
rs768122336 Chr.16: 85648686-85648686 on Build GRCh38
rs763319382 Chr.16: 85648712-85648712 on Build GRCh38
rs544831892 Chr.16: 85648662-85648662 on Build GRCh38
rs764223650 Chr.16: 85648719-85648719 on Build GRCh38
rs752961108 Chr.16: 85648704-85648704 on Build GRCh38
rs749008737 Chr.16: 85648677-85648677 on Build GRCh38
rs758340786 Chr.16: 85648602-85648602 on Build GRCh38

View Full Product Details