Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40356094-40356349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DISP2
Entrez Gene ID
85455
Gene Name
dispatched RND transporter family member 2
Gene Aliases
DISPB, HsT16908, LINC00594
UniGene
Hs.355645
Gene Chromosome Location
Chr.15: 40354359-40378755 on Build GRCh38
Transcripts
XM_011522125.2 XM_017022687.1 NM_001145643.1

Gene Symbol
PHGR1
Entrez Gene ID
644844
Gene Name
proline, histidine and glycine rich 1
Gene Aliases
-
UniGene
Hs.447537
Gene Chromosome Location
Chr.15: 40351033-40356434 on Build GRCh38
Transcripts
XM_011522125.2 XM_017022687.1 NM_001145643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6554884 c.135A>T p.P45P Chr.15: 40356189-40356189 on Build GRCh38
7256550 c.111_143del33 p.G48_P58del11 Chr.15: 40356165-40356197 on Build GRCh38
7282002 c.102_103insC p.H37fs*>47 Chr.15: 40356156-40356157 on Build GRCh38
7311584 c.76G>C p.G26R Chr.15: 40356130-40356130 on Build GRCh38
6032821 c.108C>T p.P36P Chr.15: 40356162-40356162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756707012 Chr.15: 40356197-40356198 on Build GRCh38
rs780685137 Chr.15: 40356284-40356284 on Build GRCh38
rs765226354 Chr.15: 40356131-40356132 on Build GRCh38
rs753143556 Chr.15: 40356195-40356215 on Build GRCh38
rs559450424 Chr.15: 40356249-40356249 on Build GRCh38
rs71132170 Chr.15: 40356170-40356171 on Build GRCh38
rs371153968 Chr.15: 40356195-40356195 on Build GRCh38
rs558923306 Chr.15: 40356124-40356124 on Build GRCh38
rs561449637 Chr.15: 40356216-40356216 on Build GRCh38
rs928164405 Chr.15: 40356183-40356183 on Build GRCh38
rs1011319387 Chr.15: 40356135-40356135 on Build GRCh38
rs757733858 Chr.15: 40356230-40356230 on Build GRCh38
rs747623401 Chr.15: 40356290-40356290 on Build GRCh38
rs754442317 Chr.15: 40356282-40356282 on Build GRCh38
rs777127846 Chr.15: 40356218-40356229 on Build GRCh38
rs748850271 Chr.15: 40356223-40356223 on Build GRCh38
rs12901020 Chr.15: 40356237-40356237 on Build GRCh38
rs746468153 Chr.15: 40356127-40356127 on Build GRCh38
rs759512760 Chr.15: 40356197-40356197 on Build GRCh38
rs143235232 Chr.15: 40356211-40356211 on Build GRCh38
rs776414601 Chr.15: 40356130-40356130 on Build GRCh38
rs758595423 Chr.15: 40356157-40356158 on Build GRCh38
rs1051806173 Chr.15: 40356245-40356245 on Build GRCh38
rs901699055 Chr.15: 40356252-40356252 on Build GRCh38
rs978240296 Chr.15: 40356162-40356162 on Build GRCh38
rs769726162 Chr.15: 40356163-40356163 on Build GRCh38
rs12900982 Chr.15: 40356171-40356171 on Build GRCh38
rs996107518 Chr.15: 40356266-40356266 on Build GRCh38
rs755350345 Chr.15: 40356195-40356197 on Build GRCh38
rs9707826 Chr.15: 40356204-40356204 on Build GRCh38
rs781724729 Chr.15: 40356165-40356197 on Build GRCh38
rs1049135336 Chr.15: 40356223-40356223 on Build GRCh38
rs374256648 Chr.15: 40356196-40356196 on Build GRCh38
rs528310955 Chr.15: 40356189-40356189 on Build GRCh38
rs761884807 Chr.15: 40356129-40356129 on Build GRCh38
rs1039209234 Chr.15: 40356309-40356309 on Build GRCh38
rs528206629 Chr.15: 40356272-40356272 on Build GRCh38
rs772932787 Chr.15: 40356173-40356173 on Build GRCh38
rs890700340 Chr.15: 40356268-40356268 on Build GRCh38
rs753143748 Chr.15: 40356206-40356206 on Build GRCh38
rs113856558 Chr.15: 40356139-40356139 on Build GRCh38
rs924722865 Chr.15: 40356125-40356125 on Build GRCh38
rs750572582 Chr.15: 40356139-40356171 on Build GRCh38
rs966824484 Chr.15: 40356161-40356161 on Build GRCh38
rs781675430 Chr.15: 40356194-40356195 on Build GRCh38
rs190680696 Chr.15: 40356184-40356184 on Build GRCh38
rs12903760 Chr.15: 40356270-40356270 on Build GRCh38
rs751874640 Chr.15: 40356172-40356204 on Build GRCh38
rs1050808740 Chr.15: 40356296-40356296 on Build GRCh38
rs762342101 Chr.15: 40356254-40356254 on Build GRCh38
rs542741163 Chr.15: 40356229-40356229 on Build GRCh38
rs12904995 Chr.15: 40356255-40356255 on Build GRCh38
rs35197718 Chr.15: 40356249-40356250 on Build GRCh38
rs1020002162 Chr.15: 40356160-40356160 on Build GRCh38
rs575125497 Chr.15: 40356228-40356228 on Build GRCh38
rs748182454 Chr.15: 40356131-40356131 on Build GRCh38
rs1040220220 Chr.15: 40356240-40356240 on Build GRCh38
rs991516436 Chr.15: 40356217-40356217 on Build GRCh38
rs907977842 Chr.15: 40356224-40356224 on Build GRCh38
rs768258411 Chr.15: 40356129-40356129 on Build GRCh38

View Full Product Details