Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3942058-3942301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMRK2
Entrez Gene ID
27231
Gene Name
nicotinamide riboside kinase 2
Gene Aliases
ITGB1BP3, MIBP, NRK2
UniGene
Hs.135458
Gene Chromosome Location
Chr.19: 3933103-3942416 on Build GRCh38
Transcripts
NM_001289117.1 NM_170678.2 NR_110316.1 XM_006722725.3 XM_006722726.3 XM_017026597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2152750 c.611G>A p.R204H Chr.19: 3942191-3942191 on Build GRCh38
439452 c.638G>A p.R213H Chr.19: 3942218-3942218 on Build GRCh38
6346645 c.641G>T p.G214V Chr.19: 3942221-3942221 on Build GRCh38
255634 c.545A>T p.E182V Chr.19: 3942125-3942125 on Build GRCh38
6311468 c.653G>T p.R218I Chr.19: 3942233-3942233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148078582 Chr.19: 3942218-3942218 on Build GRCh38
rs1009334959 Chr.19: 3942196-3942196 on Build GRCh38
rs144955421 Chr.19: 3942265-3942265 on Build GRCh38
rs201732259 Chr.19: 3942121-3942121 on Build GRCh38
rs150621748 Chr.19: 3942224-3942224 on Build GRCh38
rs764803188 Chr.19: 3942106-3942106 on Build GRCh38
rs754650767 Chr.19: 3942177-3942177 on Build GRCh38
rs775391418 Chr.19: 3942093-3942093 on Build GRCh38
rs753323449 Chr.19: 3942200-3942200 on Build GRCh38
rs149329333 Chr.19: 3942099-3942099 on Build GRCh38
rs776357194 Chr.19: 3942153-3942153 on Build GRCh38
rs199939061 Chr.19: 3942094-3942094 on Build GRCh38
rs768503652 Chr.19: 3942150-3942150 on Build GRCh38
rs140172193 Chr.19: 3942225-3942225 on Build GRCh38
rs568033275 Chr.19: 3942125-3942125 on Build GRCh38
rs758999064 Chr.19: 3942264-3942264 on Build GRCh38
rs1019834285 Chr.19: 3942205-3942205 on Build GRCh38
rs754402238 Chr.19: 3942236-3942236 on Build GRCh38
rs780174331 Chr.19: 3942242-3942242 on Build GRCh38
rs144739333 Chr.19: 3942209-3942209 on Build GRCh38
rs143940174 Chr.19: 3942232-3942232 on Build GRCh38
rs749419134 Chr.19: 3942132-3942132 on Build GRCh38
rs745997876 Chr.19: 3942239-3942239 on Build GRCh38
rs751883079 Chr.19: 3942118-3942118 on Build GRCh38
rs201693415 Chr.19: 3942220-3942220 on Build GRCh38
rs1055380986 Chr.19: 3942255-3942255 on Build GRCh38
rs772240738 Chr.19: 3942212-3942212 on Build GRCh38
rs765140251 Chr.19: 3942161-3942161 on Build GRCh38
rs761036295 Chr.19: 3942274-3942274 on Build GRCh38
rs138906846 Chr.19: 3942122-3942122 on Build GRCh38
rs756887862 Chr.19: 3942248-3942248 on Build GRCh38
rs771323973 Chr.19: 3942087-3942087 on Build GRCh38
rs747937794 Chr.19: 3942214-3942214 on Build GRCh38
rs141476437 Chr.19: 3942149-3942149 on Build GRCh38
rs1050830481 Chr.19: 3942227-3942227 on Build GRCh38
rs749330791 Chr.19: 3942191-3942191 on Build GRCh38
rs36093414 Chr.19: 3942180-3942180 on Build GRCh38
rs566515700 Chr.19: 3942275-3942275 on Build GRCh38
rs765956253 Chr.19: 3942174-3942174 on Build GRCh38
rs552712471 Chr.19: 3942109-3942109 on Build GRCh38
rs572917677 Chr.19: 3942198-3942198 on Build GRCh38
rs962944322 Chr.19: 3942211-3942211 on Build GRCh38
rs372708977 Chr.19: 3942276-3942276 on Build GRCh38
rs200114937 Chr.19: 3942107-3942107 on Build GRCh38
rs138618157 Chr.19: 3942098-3942098 on Build GRCh38
rs773924875 Chr.19: 3942130-3942131 on Build GRCh38
rs763249774 Chr.19: 3942083-3942083 on Build GRCh38
rs146981587 Chr.19: 3942185-3942185 on Build GRCh38
rs547115351 Chr.19: 3942252-3942252 on Build GRCh38
rs749907658 Chr.19: 3942163-3942163 on Build GRCh38
rs775016622 Chr.19: 3942269-3942269 on Build GRCh38
rs972177328 Chr.19: 3942243-3942243 on Build GRCh38
rs775465439 Chr.19: 3942103-3942103 on Build GRCh38
rs764640413 Chr.19: 3942233-3942233 on Build GRCh38
rs773818812 Chr.19: 3942256-3942256 on Build GRCh38
rs141849847 Chr.19: 3942223-3942223 on Build GRCh38
rs138158318 Chr.19: 3942197-3942197 on Build GRCh38
rs762483675 Chr.19: 3942167-3942167 on Build GRCh38
rs375051879 Chr.19: 3942207-3942207 on Build GRCh38
rs753831606 Chr.19: 3942101-3942101 on Build GRCh38
rs149893927 Chr.19: 3942251-3942251 on Build GRCh38
rs181454061 Chr.19: 3942160-3942160 on Build GRCh38
rs751214143 Chr.19: 3942176-3942176 on Build GRCh38
rs767115630 Chr.19: 3942226-3942226 on Build GRCh38
rs35701787 Chr.19: 3942247-3942247 on Build GRCh38
rs371867237 Chr.19: 3942190-3942190 on Build GRCh38
rs16992131 Chr.19: 3942112-3942112 on Build GRCh38
rs146363585 Chr.19: 3942237-3942237 on Build GRCh38
rs201205688 Chr.19: 3942138-3942138 on Build GRCh38
rs777883897 Chr.19: 3942128-3942128 on Build GRCh38
rs199529821 Chr.19: 3942186-3942186 on Build GRCh38
rs758619042 Chr.19: 3942241-3942241 on Build GRCh38
rs369195071 Chr.19: 3942217-3942217 on Build GRCh38
rs773232246 Chr.19: 3942080-3942080 on Build GRCh38
rs560264955 Chr.19: 3942213-3942213 on Build GRCh38

View Full Product Details