Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53848545-53848802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL17RB
Entrez Gene ID
55540
Gene Name
interleukin 17 receptor B
Gene Aliases
CRL4, EVI27, IL17BR, IL17RH1
UniGene
Hs.654970
Gene Chromosome Location
Chr.3: 53846494-53866526 on Build GRCh38
Transcripts
NM_018725.3 XM_005265310.4 XM_005265311.4 XM_005265312.4 XM_011533941.2 XM_017006804.1 XM_017006805.1 XM_017006806.1 XM_017006807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3945610 c.85+10T>C p.? Chr.3: 53848698-53848698 on Build GRCh38
5892877 c.63C>T p.T21T Chr.3: 53848666-53848666 on Build GRCh38
5432623 c.67C>T p.Q23* Chr.3: 53848670-53848670 on Build GRCh38
71146 c.64G>A p.V22I Chr.3: 53848667-53848667 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759986070 Chr.3: 53848706-53848706 on Build GRCh38
rs766805379 Chr.3: 53848754-53848754 on Build GRCh38
rs747457050 Chr.3: 53848677-53848677 on Build GRCh38
rs113111250 Chr.3: 53848641-53848641 on Build GRCh38
rs374807881 Chr.3: 53848626-53848626 on Build GRCh38
rs1028027958 Chr.3: 53848769-53848769 on Build GRCh38
rs768173205 Chr.3: 53848708-53848708 on Build GRCh38
rs749333729 Chr.3: 53848725-53848725 on Build GRCh38
rs751004115 Chr.3: 53848632-53848632 on Build GRCh38
rs1002658624 Chr.3: 53848590-53848590 on Build GRCh38
rs112485971 Chr.3: 53848672-53848672 on Build GRCh38
rs367947716 Chr.3: 53848722-53848722 on Build GRCh38
rs763873666 Chr.3: 53848627-53848627 on Build GRCh38
rs1031424938 Chr.3: 53848609-53848609 on Build GRCh38
rs5849009 Chr.3: 53848646-53848647 on Build GRCh38
rs397764175 Chr.3: 53848652-53848653 on Build GRCh38
rs755121971 Chr.3: 53848658-53848658 on Build GRCh38
rs200784572 Chr.3: 53848646-53848646 on Build GRCh38
rs1053830544 Chr.3: 53848604-53848604 on Build GRCh38
rs753728728 Chr.3: 53848655-53848655 on Build GRCh38
rs565147147 Chr.3: 53848767-53848767 on Build GRCh38
rs372245466 Chr.3: 53848615-53848615 on Build GRCh38
rs923512092 Chr.3: 53848703-53848703 on Build GRCh38
rs377143258 Chr.3: 53848666-53848666 on Build GRCh38
rs368270454 Chr.3: 53848644-53848645 on Build GRCh38
rs762059693 Chr.3: 53848636-53848637 on Build GRCh38
rs761199589 Chr.3: 53848617-53848617 on Build GRCh38
rs7650686 Chr.3: 53848768-53848768 on Build GRCh38
rs771485961 Chr.3: 53848687-53848687 on Build GRCh38
rs199736035 Chr.3: 53848639-53848639 on Build GRCh38
rs869279235 Chr.3: 53848653-53848654 on Build GRCh38
rs975430454 Chr.3: 53848695-53848695 on Build GRCh38
rs774009147 Chr.3: 53848720-53848720 on Build GRCh38
rs776976246 Chr.3: 53848704-53848704 on Build GRCh38
rs1025690 Chr.3: 53848636-53848636 on Build GRCh38
rs767086796 Chr.3: 53848622-53848622 on Build GRCh38
rs756129785 Chr.3: 53848640-53848640 on Build GRCh38
rs932328596 Chr.3: 53848582-53848582 on Build GRCh38
rs750348451 Chr.3: 53848638-53848638 on Build GRCh38
rs140254923 Chr.3: 53848667-53848667 on Build GRCh38
rs2232333 Chr.3: 53848634-53848634 on Build GRCh38

View Full Product Details