Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36326729-36326994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0895
Entrez Gene ID
23366
Gene Name
KIAA0895
Gene Aliases
-
UniGene
Hs.6224
Gene Chromosome Location
Chr.7: 36324150-36390125 on Build GRCh38
Transcripts
NM_001100425.1 NM_001199706.1 NM_001199707.1 NM_001199708.1 NM_001300956.1 NM_015314.2 XM_005249689.3 XM_005249690.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4812987 c.1334G>A p.G445D Chr.7: 36326836-36326836 on Build GRCh38
6109917 c.1306A>G p.R436G Chr.7: 36326864-36326864 on Build GRCh38
3880801 c.1411G>A p.E471K Chr.7: 36326873-36326873 on Build GRCh38
4809167 c.1287G>C p.L429F Chr.7: 36326883-36326883 on Build GRCh38
1719270 c.1424T>C p.V475A Chr.7: 36326860-36326860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769839417 Chr.7: 36326847-36326847 on Build GRCh38
rs758796151 Chr.7: 36326946-36326946 on Build GRCh38
rs4723513 Chr.7: 36326874-36326874 on Build GRCh38
rs781130013 Chr.7: 36326969-36326969 on Build GRCh38
rs758679663 Chr.7: 36326813-36326813 on Build GRCh38
rs767891674 Chr.7: 36326943-36326943 on Build GRCh38
rs780271725 Chr.7: 36326846-36326846 on Build GRCh38
rs867170191 Chr.7: 36326854-36326854 on Build GRCh38
rs563885993 Chr.7: 36326865-36326865 on Build GRCh38
rs772782054 Chr.7: 36326904-36326904 on Build GRCh38
rs376810223 Chr.7: 36326763-36326763 on Build GRCh38
rs751675478 Chr.7: 36326952-36326952 on Build GRCh38
rs568010970 Chr.7: 36326873-36326873 on Build GRCh38
rs778181902 Chr.7: 36326822-36326822 on Build GRCh38
rs767082685 Chr.7: 36326761-36326761 on Build GRCh38
rs749418966 Chr.7: 36326754-36326754 on Build GRCh38
rs370304999 Chr.7: 36326929-36326929 on Build GRCh38
rs196655 Chr.7: 36326960-36326960 on Build GRCh38
rs1011288983 Chr.7: 36326836-36326836 on Build GRCh38
rs747287289 Chr.7: 36326826-36326826 on Build GRCh38
rs766878717 Chr.7: 36326899-36326899 on Build GRCh38
rs755520772 Chr.7: 36326792-36326792 on Build GRCh38
rs191576648 Chr.7: 36326795-36326795 on Build GRCh38
rs548117983 Chr.7: 36326833-36326833 on Build GRCh38
rs373853280 Chr.7: 36326938-36326938 on Build GRCh38
rs542093919 Chr.7: 36326752-36326752 on Build GRCh38
rs781346763 Chr.7: 36326834-36326834 on Build GRCh38
rs376971921 Chr.7: 36326878-36326879 on Build GRCh38
rs61737930 Chr.7: 36326892-36326892 on Build GRCh38
rs753020071 Chr.7: 36326950-36326950 on Build GRCh38
rs754632958 Chr.7: 36326822-36326823 on Build GRCh38
rs373694114 Chr.7: 36326755-36326755 on Build GRCh38
rs531386040 Chr.7: 36326806-36326806 on Build GRCh38

View Full Product Details