Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49373365-49373606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNB1
Entrez Gene ID
3745
Gene Name
potassium voltage-gated channel subfamily B member 1
Gene Aliases
DRK1, Kv2.1
UniGene
Hs.633143
Gene Chromosome Location
Chr.20: 49363877-49484033 on Build GRCh38
Transcripts
NM_004975.3 XM_006723784.3 XM_011528799.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
724252 c.1963G>A p.E655K Chr.20: 49373597-49373597 on Build GRCh38
1255245 c.2092C>A p.R698R Chr.20: 49373468-49373468 on Build GRCh38
4383302 c.1990A>G p.N664D Chr.20: 49373570-49373570 on Build GRCh38
4134640 c.2106G>A p.A702A Chr.20: 49373454-49373454 on Build GRCh38
3547562 c.1997C>T p.P666L Chr.20: 49373563-49373563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776815894 Chr.20: 49373491-49373491 on Build GRCh38
rs759700203 Chr.20: 49373418-49373418 on Build GRCh38
rs139458439 Chr.20: 49373424-49373424 on Build GRCh38
rs898934503 Chr.20: 49373575-49373575 on Build GRCh38
rs779379680 Chr.20: 49373468-49373468 on Build GRCh38
rs954205763 Chr.20: 49373391-49373391 on Build GRCh38
rs747112059 Chr.20: 49373580-49373580 on Build GRCh38
rs756918855 Chr.20: 49373561-49373561 on Build GRCh38
rs771624206 Chr.20: 49373393-49373393 on Build GRCh38
rs377306621 Chr.20: 49373534-49373534 on Build GRCh38
rs779750227 Chr.20: 49373444-49373444 on Build GRCh38
rs764306025 Chr.20: 49373431-49373431 on Build GRCh38
rs752215642 Chr.20: 49373526-49373526 on Build GRCh38
rs201212125 Chr.20: 49373563-49373563 on Build GRCh38
rs753575587 Chr.20: 49373551-49373551 on Build GRCh38
rs758890482 Chr.20: 49373568-49373568 on Build GRCh38
rs751308084 Chr.20: 49373511-49373511 on Build GRCh38
rs775433680 Chr.20: 49373477-49373477 on Build GRCh38
rs749718924 Chr.20: 49373463-49373463 on Build GRCh38
rs767422124 Chr.20: 49373525-49373525 on Build GRCh38
rs772569916 Chr.20: 49373473-49373473 on Build GRCh38
rs755628300 Chr.20: 49373532-49373532 on Build GRCh38
rs765489173 Chr.20: 49373504-49373506 on Build GRCh38
rs768928325 Chr.20: 49373484-49373484 on Build GRCh38
rs908785527 Chr.20: 49373502-49373502 on Build GRCh38
rs144379782 Chr.20: 49373455-49373455 on Build GRCh38
rs571848522 Chr.20: 49373429-49373429 on Build GRCh38
rs149678923 Chr.20: 49373454-49373454 on Build GRCh38
rs771391112 Chr.20: 49373464-49373464 on Build GRCh38
rs199910304 Chr.20: 49373445-49373445 on Build GRCh38
rs746382408 Chr.20: 49373469-49373469 on Build GRCh38
rs760800348 Chr.20: 49373479-49373479 on Build GRCh38
rs746173205 Chr.20: 49373565-49373565 on Build GRCh38
rs372263564 Chr.20: 49373501-49373501 on Build GRCh38
rs780593043 Chr.20: 49373577-49373577 on Build GRCh38
rs6090970 Chr.20: 49373412-49373412 on Build GRCh38
rs762727724 Chr.20: 49373443-49373443 on Build GRCh38
rs774976039 Chr.20: 49373408-49373408 on Build GRCh38
rs759406413 Chr.20: 49373515-49373515 on Build GRCh38
rs750280554 Chr.20: 49373433-49373433 on Build GRCh38
rs777563722 Chr.20: 49373462-49373462 on Build GRCh38
rs775588653 Chr.20: 49373425-49373425 on Build GRCh38
rs200849612 Chr.20: 49373505-49373505 on Build GRCh38

View Full Product Details