Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48464245-48464476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ14
Entrez Gene ID
3770
Gene Name
potassium voltage-gated channel subfamily J member 14
Gene Aliases
IRK4, KIR2.4
UniGene
Hs.144011
Gene Chromosome Location
Chr.19: 48455707-48466110 on Build GRCh38
Transcripts
NM_013348.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
349622 c.810C>T p.S270S Chr.19: 48464276-48464276 on Build GRCh38
3536506 c.834C>T p.I278I Chr.19: 48464300-48464300 on Build GRCh38
3536507 c.842C>T p.A281V Chr.19: 48464308-48464308 on Build GRCh38
3107398 c.793C>T p.R265C Chr.19: 48464259-48464259 on Build GRCh38
4602454 c.892G>C p.E298Q Chr.19: 48464358-48464358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899566507 Chr.19: 48464395-48464395 on Build GRCh38
rs745329360 Chr.19: 48464356-48464356 on Build GRCh38
rs200963621 Chr.19: 48464425-48464425 on Build GRCh38
rs374178623 Chr.19: 48464271-48464271 on Build GRCh38
rs746854974 Chr.19: 48464373-48464373 on Build GRCh38
rs754270596 Chr.19: 48464338-48464338 on Build GRCh38
rs760874158 Chr.19: 48464409-48464410 on Build GRCh38
rs769720803 Chr.19: 48464403-48464403 on Build GRCh38
rs139400525 Chr.19: 48464280-48464280 on Build GRCh38
rs149035039 Chr.19: 48464336-48464336 on Build GRCh38
rs949961054 Chr.19: 48464297-48464297 on Build GRCh38
rs151174361 Chr.19: 48464390-48464390 on Build GRCh38
rs267605565 Chr.19: 48464300-48464300 on Build GRCh38
rs757964679 Chr.19: 48464454-48464454 on Build GRCh38
rs1032306591 Chr.19: 48464301-48464301 on Build GRCh38
rs375930558 Chr.19: 48464420-48464420 on Build GRCh38
rs762242985 Chr.19: 48464430-48464430 on Build GRCh38
rs996562660 Chr.19: 48464405-48464405 on Build GRCh38
rs887066654 Chr.19: 48464334-48464334 on Build GRCh38
rs763139395 Chr.19: 48464276-48464276 on Build GRCh38
rs562175440 Chr.19: 48464287-48464287 on Build GRCh38
rs771804173 Chr.19: 48464267-48464267 on Build GRCh38
rs754184467 Chr.19: 48464427-48464427 on Build GRCh38
rs3745725 Chr.19: 48464331-48464331 on Build GRCh38
rs767668128 Chr.19: 48464289-48464289 on Build GRCh38
rs144173583 Chr.19: 48464288-48464288 on Build GRCh38
rs779029656 Chr.19: 48464275-48464275 on Build GRCh38
rs750025228 Chr.19: 48464436-48464436 on Build GRCh38
rs765711766 Chr.19: 48464432-48464432 on Build GRCh38
rs775442705 Chr.19: 48464268-48464268 on Build GRCh38
rs143059922 Chr.19: 48464350-48464350 on Build GRCh38
rs757745215 Chr.19: 48464341-48464341 on Build GRCh38
rs772340028 Chr.19: 48464419-48464419 on Build GRCh38
rs748066542 Chr.19: 48464398-48464398 on Build GRCh38
rs766335547 Chr.19: 48464284-48464284 on Build GRCh38
rs764713358 Chr.19: 48464337-48464337 on Build GRCh38
rs768547413 Chr.19: 48464375-48464375 on Build GRCh38
rs759545604 Chr.19: 48464416-48464416 on Build GRCh38
rs779845193 Chr.19: 48464366-48464366 on Build GRCh38
rs371264426 Chr.19: 48464363-48464363 on Build GRCh38
rs373780901 Chr.19: 48464391-48464392 on Build GRCh38
rs774308144 Chr.19: 48464415-48464415 on Build GRCh38
rs146736268 Chr.19: 48464270-48464270 on Build GRCh38

View Full Product Details