Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 41947406-41947663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf91
Entrez Gene ID
400950
Gene Name
chromosome 2 open reading frame 91
Gene Aliases
-
UniGene
Hs.738713
Gene Chromosome Location
Chr.2: 41935368-41961382 on Build GRCh38
Transcripts
NM_001242815.1 XM_017004097.1

SNP Detail

SNP ID Chromosome Location
rs775314029 Chr.2: 41947481-41947481 on Build GRCh38
rs949523370 Chr.2: 41947519-41947519 on Build GRCh38
rs553440986 Chr.2: 41947558-41947558 on Build GRCh38
rs771081024 Chr.2: 41947531-41947531 on Build GRCh38
rs763588153 Chr.2: 41947562-41947562 on Build GRCh38
rs896565628 Chr.2: 41947472-41947472 on Build GRCh38
rs763748762 Chr.2: 41947542-41947542 on Build GRCh38
rs879693359 Chr.2: 41947441-41947441 on Build GRCh38
rs935341719 Chr.2: 41947425-41947425 on Build GRCh38
rs865895427 Chr.2: 41947451-41947451 on Build GRCh38
rs1033030911 Chr.2: 41947608-41947608 on Build GRCh38
rs771945465 Chr.2: 41947468-41947468 on Build GRCh38
rs548542030 Chr.2: 41947627-41947627 on Build GRCh38
rs749403675 Chr.2: 41947484-41947484 on Build GRCh38
rs1006116378 Chr.2: 41947638-41947638 on Build GRCh38
rs760376479 Chr.2: 41947498-41947498 on Build GRCh38
rs1044890760 Chr.2: 41947587-41947587 on Build GRCh38
rs115037838 Chr.2: 41947537-41947537 on Build GRCh38
rs775995830 Chr.2: 41947549-41947549 on Build GRCh38
rs144307729 Chr.2: 41947474-41947474 on Build GRCh38
rs140290330 Chr.2: 41947466-41947466 on Build GRCh38
rs983833102 Chr.2: 41947460-41947460 on Build GRCh38
rs28548998 Chr.2: 41947593-41947593 on Build GRCh38
rs766806010 Chr.2: 41947568-41947568 on Build GRCh38
rs755364742 Chr.2: 41947595-41947595 on Build GRCh38
rs746151117 Chr.2: 41947634-41947634 on Build GRCh38
rs201863817 Chr.2: 41947430-41947430 on Build GRCh38
rs878950967 Chr.2: 41947640-41947640 on Build GRCh38
rs909641747 Chr.2: 41947462-41947462 on Build GRCh38

View Full Product Details