Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37951363-37951616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf23
Entrez Gene ID
84645
Gene Name
chromosome 22 open reading frame 23
Gene Aliases
EVG1, dJ1039K5.6
UniGene
Hs.517612
Gene Chromosome Location
Chr.22: 37943050-37953669 on Build GRCh38
Transcripts
NM_001207062.1 NM_032561.4 XM_005261781.1 XM_005261782.3 XM_005261783.2 XM_005261784.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
478957 c.104-1G>A p.? Chr.22: 37951523-37951523 on Build GRCh38
6615664 c.144C>T p.R48R Chr.22: 37951482-37951482 on Build GRCh38
4582397 c.154G>T p.D52Y Chr.22: 37951472-37951472 on Build GRCh38
5633964 c.150C>G p.I50M Chr.22: 37951476-37951476 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs935217971 Chr.22: 37951542-37951542 on Build GRCh38
rs934583107 Chr.22: 37951409-37951409 on Build GRCh38
rs756698757 Chr.22: 37951466-37951466 on Build GRCh38
rs777362581 Chr.22: 37951465-37951465 on Build GRCh38
rs377650163 Chr.22: 37951498-37951498 on Build GRCh38
rs762703519 Chr.22: 37951556-37951556 on Build GRCh38
rs369688391 Chr.22: 37951564-37951564 on Build GRCh38
rs114523199 Chr.22: 37951483-37951483 on Build GRCh38
rs772960439 Chr.22: 37951514-37951516 on Build GRCh38
rs763784937 Chr.22: 37951563-37951563 on Build GRCh38
rs369094019 Chr.22: 37951457-37951457 on Build GRCh38
rs752433207 Chr.22: 37951448-37951448 on Build GRCh38
rs765113011 Chr.22: 37951533-37951533 on Build GRCh38
rs200370493 Chr.22: 37951444-37951444 on Build GRCh38
rs760643124 Chr.22: 37951423-37951423 on Build GRCh38
rs752518890 Chr.22: 37951543-37951543 on Build GRCh38
rs766391851 Chr.22: 37951433-37951433 on Build GRCh38
rs563655560 Chr.22: 37951584-37951608 on Build GRCh38
rs975350506 Chr.22: 37951470-37951470 on Build GRCh38
rs759406418 Chr.22: 37951529-37951529 on Build GRCh38
rs749750122 Chr.22: 37951476-37951476 on Build GRCh38
rs141612323 Chr.22: 37951484-37951484 on Build GRCh38
rs764947485 Chr.22: 37951447-37951447 on Build GRCh38
rs146937811 Chr.22: 37951475-37951475 on Build GRCh38
rs9610883 Chr.22: 37951540-37951540 on Build GRCh38
rs772171983 Chr.22: 37951479-37951490 on Build GRCh38
rs767787441 Chr.22: 37951412-37951412 on Build GRCh38
rs77071478 Chr.22: 37951455-37951455 on Build GRCh38
rs201828953 Chr.22: 37951442-37951442 on Build GRCh38
rs12170918 Chr.22: 37951497-37951497 on Build GRCh38
rs951605583 Chr.22: 37951440-37951440 on Build GRCh38
rs371739699 Chr.22: 37951468-37951468 on Build GRCh38
rs773459398 Chr.22: 37951418-37951418 on Build GRCh38
rs751052783 Chr.22: 37951548-37951548 on Build GRCh38
rs188271512 Chr.22: 37951587-37951587 on Build GRCh38
rs150911308 Chr.22: 37951493-37951493 on Build GRCh38
rs776707271 Chr.22: 37951523-37951523 on Build GRCh38
rs762128726 Chr.22: 37951411-37951411 on Build GRCh38
rs766365932 Chr.22: 37951512-37951512 on Build GRCh38

View Full Product Details