Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87416472-87416745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP206
Entrez Gene ID
154313
Gene Name
cilia and flagella associated protein 206
Gene Aliases
C6orf165, dJ382I10.1
UniGene
Hs.82921
Gene Chromosome Location
Chr.6: 87407972-87464473 on Build GRCh38
Transcripts
NM_001031743.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5618290 c.509G>C p.G170A Chr.6: 87416705-87416705 on Build GRCh38
159642 c.502G>C p.E168Q Chr.6: 87416698-87416698 on Build GRCh38
6888523 c.534G>A p.K178K Chr.6: 87416730-87416730 on Build GRCh38
5618291 c.509G>C p.G170A Chr.6: 87416705-87416705 on Build GRCh38
3413955 c.544C>T p.R182C Chr.6: 87416740-87416740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758240197 Chr.6: 87416655-87416655 on Build GRCh38
rs973726469 Chr.6: 87416560-87416560 on Build GRCh38
rs760849444 Chr.6: 87416710-87416710 on Build GRCh38
rs184967058 Chr.6: 87416653-87416653 on Build GRCh38
rs753422473 Chr.6: 87416678-87416679 on Build GRCh38
rs369303324 Chr.6: 87416657-87416657 on Build GRCh38
rs778470371 Chr.6: 87416690-87416690 on Build GRCh38
rs76159319 Chr.6: 87416654-87416654 on Build GRCh38
rs559398329 Chr.6: 87416660-87416660 on Build GRCh38
rs776015267 Chr.6: 87416624-87416624 on Build GRCh38
rs748644428 Chr.6: 87416678-87416678 on Build GRCh38
rs542071458 Chr.6: 87416708-87416708 on Build GRCh38
rs750190343 Chr.6: 87416632-87416632 on Build GRCh38
rs962990121 Chr.6: 87416545-87416545 on Build GRCh38
rs139163914 Chr.6: 87416609-87416610 on Build GRCh38
rs985726468 Chr.6: 87416505-87416505 on Build GRCh38
rs16879281 Chr.6: 87416705-87416705 on Build GRCh38
rs775159673 Chr.6: 87416643-87416643 on Build GRCh38
rs753751096 Chr.6: 87416618-87416618 on Build GRCh38
rs1036526879 Chr.6: 87416663-87416663 on Build GRCh38
rs774646874 Chr.6: 87416704-87416704 on Build GRCh38
rs929685480 Chr.6: 87416577-87416577 on Build GRCh38
rs771487554 Chr.6: 87416697-87416697 on Build GRCh38
rs960823481 Chr.6: 87416562-87416563 on Build GRCh38
rs770165847 Chr.6: 87416684-87416684 on Build GRCh38
rs750419478 Chr.6: 87416718-87416718 on Build GRCh38
rs759167400 Chr.6: 87416718-87416721 on Build GRCh38
rs1018184524 Chr.6: 87416529-87416529 on Build GRCh38
rs761676970 Chr.6: 87416620-87416620 on Build GRCh38
rs752233050 Chr.6: 87416610-87416610 on Build GRCh38
rs780584317 Chr.6: 87416676-87416676 on Build GRCh38
rs992238861 Chr.6: 87416594-87416594 on Build GRCh38
rs112853646 Chr.6: 87416662-87416662 on Build GRCh38
rs775464503 Chr.6: 87416709-87416709 on Build GRCh38
rs763448560 Chr.6: 87416636-87416636 on Build GRCh38
rs969810853 Chr.6: 87416609-87416609 on Build GRCh38
rs967292536 Chr.6: 87416508-87416508 on Build GRCh38
rs998718453 Chr.6: 87416521-87416521 on Build GRCh38
rs765042682 Chr.6: 87416631-87416631 on Build GRCh38
rs918266031 Chr.6: 87416566-87416566 on Build GRCh38
rs1035505189 Chr.6: 87416555-87416555 on Build GRCh38
rs533809890 Chr.6: 87416504-87416504 on Build GRCh38
rs181724340 Chr.6: 87416497-87416497 on Build GRCh38
rs529616908 Chr.6: 87416642-87416643 on Build GRCh38
rs369501687 Chr.6: 87416601-87416601 on Build GRCh38
rs760384345 Chr.6: 87416643-87416644 on Build GRCh38
rs749653642 Chr.6: 87416692-87416692 on Build GRCh38

View Full Product Details