Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119232485-119232734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBL
Entrez Gene ID
867
Gene Name
Cbl proto-oncogene
Gene Aliases
C-CBL, CBL2, FRA11B, NSLL, RNF55
UniGene
Hs.504096
Gene Chromosome Location
Chr.11: 119206276-119308149 on Build GRCh38
Transcripts
NM_005188.3 XM_011543057.2 XM_017018511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6438112 c.299C>G p.S100* Chr.11: 119232551-119232551 on Build GRCh38
923809 c.279G>A p.Q93Q Chr.11: 119232531-119232531 on Build GRCh38
391718 c.352delT p.F118fs*5 Chr.11: 119232604-119232604 on Build GRCh38
6960300 c.422A>G p.Y141C Chr.11: 119232674-119232674 on Build GRCh38
6563972 c.358G>C p.E120Q Chr.11: 119232610-119232610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147438359 Chr.11: 119232538-119232538 on Build GRCh38
rs763336179 Chr.11: 119232648-119232648 on Build GRCh38
rs755204320 Chr.11: 119232690-119232690 on Build GRCh38
rs768782468 Chr.11: 119232665-119232666 on Build GRCh38
rs770209522 Chr.11: 119232637-119232637 on Build GRCh38
rs587778160 Chr.11: 119232523-119232523 on Build GRCh38
rs368959076 Chr.11: 119232696-119232696 on Build GRCh38
rs397507489 Chr.11: 119232558-119232558 on Build GRCh38
rs1026372456 Chr.11: 119232570-119232570 on Build GRCh38
rs145011318 Chr.11: 119232660-119232660 on Build GRCh38
rs774626378 Chr.11: 119232667-119232667 on Build GRCh38
rs761445733 Chr.11: 119232702-119232702 on Build GRCh38
rs139139107 Chr.11: 119232553-119232553 on Build GRCh38
rs760633685 Chr.11: 119232689-119232689 on Build GRCh38
rs748412298 Chr.11: 119232599-119232599 on Build GRCh38
rs1057519030 Chr.11: 119232657-119232657 on Build GRCh38
rs587778161 Chr.11: 119232640-119232640 on Build GRCh38
rs868705826 Chr.11: 119232634-119232634 on Build GRCh38
rs781738309 Chr.11: 119232539-119232539 on Build GRCh38
rs753823608 Chr.11: 119232701-119232701 on Build GRCh38
rs773502403 Chr.11: 119232645-119232645 on Build GRCh38

View Full Product Details