Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74523462-74523721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300LB
Entrez Gene ID
124599
Gene Name
CD300 molecule like family member b
Gene Aliases
CD300b, CLM-7, CLM7, CMRF35-A2, IREM-3, IREM3, TREM-5, TREM5
UniGene
Hs.313343
Gene Chromosome Location
Chr.17: 74521174-74531474 on Build GRCh38
Transcripts
NM_174892.3 XM_005257027.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3932720 c.482-1G>C p.? Chr.17: 74523652-74523652 on Build GRCh38
5972395 c.527T>C p.M176T Chr.17: 74523606-74523606 on Build GRCh38
394165 c.519C>A p.N173K Chr.17: 74523614-74523614 on Build GRCh38
4415940 c.482-8G>A p.? Chr.17: 74523659-74523659 on Build GRCh38
5722677 c.509C>T p.S170L Chr.17: 74523624-74523624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137895820 Chr.17: 74523629-74523629 on Build GRCh38
rs61162320 Chr.17: 74523499-74523499 on Build GRCh38
rs779617250 Chr.17: 74523634-74523634 on Build GRCh38
rs142435917 Chr.17: 74523645-74523645 on Build GRCh38
rs901149072 Chr.17: 74523500-74523500 on Build GRCh38
rs372672106 Chr.17: 74523699-74523699 on Build GRCh38
rs143428568 Chr.17: 74523616-74523616 on Build GRCh38
rs762350959 Chr.17: 74523614-74523614 on Build GRCh38
rs764543166 Chr.17: 74523606-74523606 on Build GRCh38
rs760991369 Chr.17: 74523671-74523671 on Build GRCh38
rs370650884 Chr.17: 74523703-74523703 on Build GRCh38
rs780940093 Chr.17: 74523547-74523547 on Build GRCh38
rs146364730 Chr.17: 74523676-74523676 on Build GRCh38
rs748147677 Chr.17: 74523549-74523549 on Build GRCh38
rs765802456 Chr.17: 74523691-74523691 on Build GRCh38
rs769906446 Chr.17: 74523658-74523658 on Build GRCh38
rs1472726 Chr.17: 74523659-74523659 on Build GRCh38
rs202114723 Chr.17: 74523682-74523682 on Build GRCh38
rs57288996 Chr.17: 74523484-74523484 on Build GRCh38
rs776061321 Chr.17: 74523551-74523551 on Build GRCh38
rs759815150 Chr.17: 74523578-74523578 on Build GRCh38
rs754687733 Chr.17: 74523637-74523637 on Build GRCh38
rs772288726 Chr.17: 74523661-74523661 on Build GRCh38
rs751245293 Chr.17: 74523635-74523635 on Build GRCh38
rs372167958 Chr.17: 74523544-74523544 on Build GRCh38
rs375010301 Chr.17: 74523570-74523570 on Build GRCh38
rs367952780 Chr.17: 74523612-74523612 on Build GRCh38
rs781081246 Chr.17: 74523639-74523639 on Build GRCh38
rs868327740 Chr.17: 74523591-74523591 on Build GRCh38
rs779813963 Chr.17: 74523532-74523532 on Build GRCh38
rs760913699 Chr.17: 74523679-74523679 on Build GRCh38
rs773714408 Chr.17: 74523702-74523702 on Build GRCh38
rs760893352 Chr.17: 74523596-74523596 on Build GRCh38
rs557624854 Chr.17: 74523520-74523520 on Build GRCh38
rs764705121 Chr.17: 74523615-74523615 on Build GRCh38
rs745874819 Chr.17: 74523660-74523660 on Build GRCh38
rs771103106 Chr.17: 74523554-74523554 on Build GRCh38
rs888950537 Chr.17: 74523491-74523491 on Build GRCh38
rs768874346 Chr.17: 74523674-74523674 on Build GRCh38
rs370309053 Chr.17: 74523538-74523538 on Build GRCh38
rs770469135 Chr.17: 74523486-74523486 on Build GRCh38
rs775529760 Chr.17: 74523668-74523668 on Build GRCh38
rs772487384 Chr.17: 74523581-74523581 on Build GRCh38
rs1038060087 Chr.17: 74523593-74523593 on Build GRCh38
rs6501721 Chr.17: 74523497-74523497 on Build GRCh38
rs994193869 Chr.17: 74523516-74523516 on Build GRCh38
rs1023360659 Chr.17: 74523644-74523644 on Build GRCh38
rs200261260 Chr.17: 74523592-74523592 on Build GRCh38

View Full Product Details