Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95565799-95566072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSK
Entrez Gene ID
153642
Gene Name
arylsulfatase family member K
Gene Aliases
TSULF
UniGene
Hs.585051
Gene Chromosome Location
Chr.5: 95555108-95605102 on Build GRCh38
Transcripts
NM_198150.2 XM_005271904.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3856961 c.196C>T p.R66C Chr.5: 95566067-95566067 on Build GRCh38
1183686 c.157C>T p.Q53* Chr.5: 95566028-95566028 on Build GRCh38
3856962 c.197G>A p.R66H Chr.5: 95566068-95566068 on Build GRCh38
3141701 c.130G>A p.G44R Chr.5: 95566001-95566001 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751588506 Chr.5: 95565996-95565996 on Build GRCh38
rs750762679 Chr.5: 95566006-95566006 on Build GRCh38
rs1056524205 Chr.5: 95566011-95566011 on Build GRCh38
rs756153393 Chr.5: 95565962-95565962 on Build GRCh38
rs781298663 Chr.5: 95565830-95565830 on Build GRCh38
rs190706511 Chr.5: 95565924-95565924 on Build GRCh38
rs964859750 Chr.5: 95565890-95565890 on Build GRCh38
rs1001635886 Chr.5: 95565882-95565882 on Build GRCh38
rs911732126 Chr.5: 95565879-95565879 on Build GRCh38
rs371079038 Chr.5: 95566044-95566044 on Build GRCh38
rs569542292 Chr.5: 95565861-95565861 on Build GRCh38
rs180767468 Chr.5: 95565970-95565970 on Build GRCh38
rs1014871383 Chr.5: 95565975-95565975 on Build GRCh38
rs974703721 Chr.5: 95565915-95565915 on Build GRCh38
rs377076352 Chr.5: 95565999-95565999 on Build GRCh38
rs892035098 Chr.5: 95565834-95565834 on Build GRCh38
rs756555588 Chr.5: 95566013-95566013 on Build GRCh38
rs749853788 Chr.5: 95566036-95566036 on Build GRCh38
rs895335030 Chr.5: 95566038-95566038 on Build GRCh38
rs1024449254 Chr.5: 95565852-95565852 on Build GRCh38
rs374433595 Chr.5: 95565995-95565995 on Build GRCh38
rs529691889 Chr.5: 95565833-95565833 on Build GRCh38
rs539875635 Chr.5: 95565964-95565964 on Build GRCh38
rs756890718 Chr.5: 95566011-95566012 on Build GRCh38
rs776696965 Chr.5: 95565959-95565959 on Build GRCh38
rs549514031 Chr.5: 95565841-95565841 on Build GRCh38
rs905999647 Chr.5: 95565871-95565871 on Build GRCh38
rs921795321 Chr.5: 95566034-95566034 on Build GRCh38
rs80012555 Chr.5: 95565951-95565951 on Build GRCh38
rs987172499 Chr.5: 95565839-95565839 on Build GRCh38
rs753619415 Chr.5: 95566009-95566010 on Build GRCh38
rs760460264 Chr.5: 95565947-95565947 on Build GRCh38
rs780254908 Chr.5: 95566035-95566035 on Build GRCh38
rs1014398751 Chr.5: 95565848-95565848 on Build GRCh38
rs767709967 Chr.5: 95566005-95566005 on Build GRCh38
rs923931074 Chr.5: 95565978-95565978 on Build GRCh38

View Full Product Details