Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152888090-152888347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFIP1
Entrez Gene ID
27236
Gene Name
ADP ribosylation factor interacting protein 1
Gene Aliases
HSU52521
UniGene
Hs.416089
Gene Chromosome Location
Chr.4: 152779937-152911912 on Build GRCh38
Transcripts
NM_001025593.2 NM_001025595.2 NM_001287431.1 NM_001287432.1 NM_001287433.1 NM_014447.3 XM_011531868.2 XM_011531869.2 XM_011531870.2 XM_011531871.2 XM_011531872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6894506 c.888C>G p.L296L Chr.4: 152888229-152888229 on Build GRCh38
1427941 c.905delA p.K303fs*13 Chr.4: 152888246-152888246 on Build GRCh38
3600959 c.952C>T p.L318L Chr.4: 152888293-152888293 on Build GRCh38
5415384 c.833A>G p.N278S Chr.4: 152888174-152888174 on Build GRCh38
3409093 c.955G>A p.E319K Chr.4: 152888296-152888296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201351020 Chr.4: 152888145-152888145 on Build GRCh38
rs142894062 Chr.4: 152888135-152888135 on Build GRCh38
rs986458954 Chr.4: 152888293-152888293 on Build GRCh38
rs752375327 Chr.4: 152888191-152888191 on Build GRCh38
rs753401929 Chr.4: 152888195-152888195 on Build GRCh38
rs767295364 Chr.4: 152888310-152888310 on Build GRCh38
rs942598070 Chr.4: 152888211-152888211 on Build GRCh38
rs972103124 Chr.4: 152888153-152888153 on Build GRCh38
rs761197917 Chr.4: 152888198-152888198 on Build GRCh38
rs367792760 Chr.4: 152888263-152888263 on Build GRCh38
rs779353354 Chr.4: 152888120-152888120 on Build GRCh38
rs758754009 Chr.4: 152888246-152888246 on Build GRCh38
rs141136069 Chr.4: 152888267-152888267 on Build GRCh38
rs766318867 Chr.4: 152888170-152888170 on Build GRCh38
rs76726136 Chr.4: 152888190-152888190 on Build GRCh38
rs755745734 Chr.4: 152888192-152888192 on Build GRCh38
rs746425395 Chr.4: 152888212-152888212 on Build GRCh38
rs776711621 Chr.4: 152888152-152888152 on Build GRCh38
rs768695550 Chr.4: 152888151-152888151 on Build GRCh38
rs564750261 Chr.4: 152888283-152888283 on Build GRCh38
rs375152331 Chr.4: 152888205-152888205 on Build GRCh38
rs368661898 Chr.4: 152888315-152888315 on Build GRCh38
rs546424412 Chr.4: 152888256-152888256 on Build GRCh38
rs747232965 Chr.4: 152888150-152888150 on Build GRCh38
rs148993045 Chr.4: 152888172-152888172 on Build GRCh38
rs748217457 Chr.4: 152888275-152888275 on Build GRCh38
rs1030779383 Chr.4: 152888168-152888168 on Build GRCh38
rs774315151 Chr.4: 152888287-152888287 on Build GRCh38
rs368185805 Chr.4: 152888185-152888185 on Build GRCh38
rs759441054 Chr.4: 152888306-152888306 on Build GRCh38
rs762813888 Chr.4: 152888162-152888162 on Build GRCh38
rs148176122 Chr.4: 152888266-152888266 on Build GRCh38
rs370704877 Chr.4: 152888193-152888193 on Build GRCh38
rs746254973 Chr.4: 152888128-152888128 on Build GRCh38
rs746459756 Chr.4: 152888181-152888181 on Build GRCh38

View Full Product Details