Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19924373-19924598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP10
Entrez Gene ID
11216
Gene Name
A-kinase anchoring protein 10
Gene Aliases
AKAP-10, D-AKAP-2, D-AKAP2, PRKA10
UniGene
Hs.642676
Gene Chromosome Location
Chr.17: 19904452-19977856 on Build GRCh38
Transcripts
NM_007202.3 XM_006721432.3 XM_017024071.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6333191 c.1706C>A p.A569E Chr.17: 19924453-19924453 on Build GRCh38
5962100 c.1699G>T p.V567L Chr.17: 19924460-19924460 on Build GRCh38
4064696 c.1692G>C p.A564A Chr.17: 19924467-19924467 on Build GRCh38
4064697 c.1656A>G p.K552K Chr.17: 19924503-19924503 on Build GRCh38
6203127 c.1690G>T p.A564S Chr.17: 19924469-19924469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772936898 Chr.17: 19924512-19924512 on Build GRCh38
rs752828043 Chr.17: 19924556-19924556 on Build GRCh38
rs776479156 Chr.17: 19924526-19924526 on Build GRCh38
rs759536868 Chr.17: 19924541-19924541 on Build GRCh38
rs748801554 Chr.17: 19924467-19924467 on Build GRCh38
rs371370941 Chr.17: 19924453-19924453 on Build GRCh38
rs983103586 Chr.17: 19924516-19924516 on Build GRCh38
rs781615376 Chr.17: 19924466-19924466 on Build GRCh38
rs142084873 Chr.17: 19924468-19924468 on Build GRCh38
rs771720764 Chr.17: 19924509-19924509 on Build GRCh38
rs969807278 Chr.17: 19924470-19924470 on Build GRCh38
rs865801175 Chr.17: 19924411-19924411 on Build GRCh38
rs151139687 Chr.17: 19924514-19924514 on Build GRCh38
rs374765026 Chr.17: 19924524-19924524 on Build GRCh38
rs201881580 Chr.17: 19924428-19924428 on Build GRCh38
rs765187438 Chr.17: 19924555-19924555 on Build GRCh38
rs921775970 Chr.17: 19924559-19924559 on Build GRCh38
rs773875750 Chr.17: 19924473-19924473 on Build GRCh38
rs201284951 Chr.17: 19924410-19924410 on Build GRCh38
rs775988115 Chr.17: 19924526-19924527 on Build GRCh38
rs1032348955 Chr.17: 19924438-19924438 on Build GRCh38
rs1005795659 Chr.17: 19924451-19924451 on Build GRCh38
rs59146308 Chr.17: 19924508-19924508 on Build GRCh38
rs924426627 Chr.17: 19924538-19924538 on Build GRCh38
rs199551208 Chr.17: 19924455-19924455 on Build GRCh38
rs372600492 Chr.17: 19924546-19924546 on Build GRCh38
rs199875653 Chr.17: 19924452-19924452 on Build GRCh38
rs375162074 Chr.17: 19924487-19924487 on Build GRCh38

View Full Product Details