Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2815196-2815457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf141
Entrez Gene ID
128653
Gene Name
chromosome 20 open reading frame 141
Gene Aliases
dJ860F19.4
UniGene
Hs.352187
Gene Chromosome Location
Chr.20: 2814987-2815830 on Build GRCh38
Transcripts
NM_001256538.1 NM_080739.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5623227 c.200C>A p.T67N Chr.20: 2815384-2815384 on Build GRCh38
4135248 c.71G>A p.G24D Chr.20: 2815255-2815255 on Build GRCh38
5809587 c.35G>T p.R12L Chr.20: 2815219-2815219 on Build GRCh38
3733630 c.23G>A p.R8K Chr.20: 2815207-2815207 on Build GRCh38
3963439 c.27C>G p.P9P Chr.20: 2815211-2815211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758291096 Chr.20: 2815419-2815419 on Build GRCh38
rs542384944 Chr.20: 2815309-2815309 on Build GRCh38
rs781064368 Chr.20: 2815340-2815340 on Build GRCh38
rs765507987 Chr.20: 2815357-2815360 on Build GRCh38
rs1009838103 Chr.20: 2815391-2815391 on Build GRCh38
rs779648219 Chr.20: 2815242-2815242 on Build GRCh38
rs140149148 Chr.20: 2815218-2815218 on Build GRCh38
rs749954548 Chr.20: 2815303-2815303 on Build GRCh38
rs754740354 Chr.20: 2815318-2815318 on Build GRCh38
rs778804020 Chr.20: 2815264-2815264 on Build GRCh38
rs759239469 Chr.20: 2815405-2815405 on Build GRCh38
rs749464395 Chr.20: 2815436-2815436 on Build GRCh38
rs755806399 Chr.20: 2815234-2815234 on Build GRCh38
rs753486981 Chr.20: 2815313-2815313 on Build GRCh38
rs749560600 Chr.20: 2815350-2815350 on Build GRCh38
rs748066707 Chr.20: 2815229-2815229 on Build GRCh38
rs147238709 Chr.20: 2815326-2815326 on Build GRCh38
rs755257504 Chr.20: 2815437-2815437 on Build GRCh38
rs764113592 Chr.20: 2815426-2815426 on Build GRCh38
rs763301245 Chr.20: 2815215-2815215 on Build GRCh38
rs142369051 Chr.20: 2815411-2815411 on Build GRCh38
rs374163831 Chr.20: 2815285-2815285 on Build GRCh38
rs12625619 Chr.20: 2815361-2815361 on Build GRCh38
rs772552150 Chr.20: 2815373-2815373 on Build GRCh38
rs919827097 Chr.20: 2815268-2815268 on Build GRCh38
rs1013252803 Chr.20: 2815314-2815314 on Build GRCh38
rs780300645 Chr.20: 2815349-2815349 on Build GRCh38
rs764950649 Chr.20: 2815407-2815407 on Build GRCh38
rs763432652 Chr.20: 2815284-2815284 on Build GRCh38
rs373769277 Chr.20: 2815385-2815385 on Build GRCh38
rs756187611 Chr.20: 2815434-2815434 on Build GRCh38
rs762468233 Chr.20: 2815289-2815289 on Build GRCh38
rs751499335 Chr.20: 2815431-2815431 on Build GRCh38
rs774869772 Chr.20: 2815355-2815355 on Build GRCh38
rs770058169 Chr.20: 2815280-2815280 on Build GRCh38
rs1021100168 Chr.20: 2815396-2815396 on Build GRCh38
rs745990496 Chr.20: 2815346-2815346 on Build GRCh38
rs371152917 Chr.20: 2815246-2815246 on Build GRCh38
rs775675120 Chr.20: 2815282-2815282 on Build GRCh38
rs954861800 Chr.20: 2815406-2815406 on Build GRCh38
rs765851219 Chr.20: 2815383-2815383 on Build GRCh38
rs529897750 Chr.20: 2815265-2815265 on Build GRCh38
rs756804548 Chr.20: 2815221-2815221 on Build GRCh38
rs770991057 Chr.20: 2815266-2815266 on Build GRCh38
rs769858186 Chr.20: 2815348-2815348 on Build GRCh38
rs752613186 Chr.20: 2815333-2815333 on Build GRCh38
rs758342612 Chr.20: 2815335-2815335 on Build GRCh38
rs1046855874 Chr.20: 2815290-2815290 on Build GRCh38
rs778499041 Chr.20: 2815228-2815228 on Build GRCh38
rs781387739 Chr.20: 2815271-2815271 on Build GRCh38
rs746016551 Chr.20: 2815274-2815274 on Build GRCh38
rs769130162 Chr.20: 2815354-2815354 on Build GRCh38
rs143838675 Chr.20: 2815219-2815219 on Build GRCh38
rs754863190 Chr.20: 2815250-2815250 on Build GRCh38
rs377607985 Chr.20: 2815292-2815292 on Build GRCh38
rs1018100039 Chr.20: 2815420-2815420 on Build GRCh38
rs552625562 Chr.20: 2815369-2815369 on Build GRCh38
rs760207763 Chr.20: 2815305-2815305 on Build GRCh38
rs757686904 Chr.20: 2815269-2815269 on Build GRCh38
rs117426258 Chr.20: 2815435-2815435 on Build GRCh38

View Full Product Details