Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150261412-150261685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA14
Entrez Gene ID
23632
Gene Name
carbonic anhydrase 14
Gene Aliases
CAXiV
UniGene
Hs.528988
Gene Chromosome Location
Chr.1: 150257255-150265078 on Build GRCh38
Transcripts
NM_012113.2 XM_005245059.2 XM_005245060.2 XM_006711259.2 XM_006711261.2 XM_011509379.2 XM_017000893.1 XM_017000894.1 XM_017000895.1 XM_017000896.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4021941 c.188C>T p.A63V Chr.1: 150261570-150261570 on Build GRCh38
4021942 c.235C>T p.L79L Chr.1: 150261617-150261617 on Build GRCh38
279505 c.223G>A p.E75K Chr.1: 150261605-150261605 on Build GRCh38
4021940 c.164T>G p.V55G Chr.1: 150261546-150261546 on Build GRCh38
4004798 c.98G>A p.W33* Chr.1: 150261480-150261480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782183375 Chr.1: 150261449-150261449 on Build GRCh38
rs916860831 Chr.1: 150261508-150261508 on Build GRCh38
rs781783234 Chr.1: 150261510-150261510 on Build GRCh38
rs782586866 Chr.1: 150261562-150261562 on Build GRCh38
rs587627022 Chr.1: 150261529-150261529 on Build GRCh38
rs781985423 Chr.1: 150261593-150261593 on Build GRCh38
rs139115533 Chr.1: 150261629-150261629 on Build GRCh38
rs782621491 Chr.1: 150261646-150261646 on Build GRCh38
rs782536133 Chr.1: 150261526-150261526 on Build GRCh38
rs373006346 Chr.1: 150261604-150261604 on Build GRCh38
rs782710442 Chr.1: 150261615-150261615 on Build GRCh38
rs782042950 Chr.1: 150261608-150261608 on Build GRCh38
rs587737167 Chr.1: 150261519-150261519 on Build GRCh38
rs369995029 Chr.1: 150261560-150261560 on Build GRCh38
rs368343263 Chr.1: 150261431-150261431 on Build GRCh38
rs971208974 Chr.1: 150261532-150261532 on Build GRCh38
rs782447059 Chr.1: 150261515-150261515 on Build GRCh38
rs782263337 Chr.1: 150261437-150261437 on Build GRCh38
rs782234596 Chr.1: 150261530-150261530 on Build GRCh38
rs924140114 Chr.1: 150261586-150261586 on Build GRCh38
rs782023468 Chr.1: 150261458-150261458 on Build GRCh38
rs782280003 Chr.1: 150261584-150261584 on Build GRCh38
rs200059954 Chr.1: 150261442-150261442 on Build GRCh38
rs782784174 Chr.1: 150261632-150261632 on Build GRCh38
rs1053855690 Chr.1: 150261616-150261616 on Build GRCh38
rs782753065 Chr.1: 150261500-150261500 on Build GRCh38
rs781966502 Chr.1: 150261605-150261605 on Build GRCh38
rs587674595 Chr.1: 150261465-150261465 on Build GRCh38
rs781831567 Chr.1: 150261635-150261635 on Build GRCh38
rs781999303 Chr.1: 150261482-150261482 on Build GRCh38
rs781864244 Chr.1: 150261520-150261520 on Build GRCh38
rs1027555162 Chr.1: 150261456-150261456 on Build GRCh38
rs782283497 Chr.1: 150261583-150261583 on Build GRCh38
rs958850147 Chr.1: 150261478-150261478 on Build GRCh38
rs373837886 Chr.1: 150261552-150261552 on Build GRCh38
rs782331580 Chr.1: 150261457-150261457 on Build GRCh38
rs112731733 Chr.1: 150261542-150261542 on Build GRCh38
rs782079968 Chr.1: 150261486-150261486 on Build GRCh38
rs782749968 Chr.1: 150261470-150261470 on Build GRCh38
rs782255287 Chr.1: 150261594-150261594 on Build GRCh38
rs782011977 Chr.1: 150261627-150261627 on Build GRCh38
rs376697011 Chr.1: 150261444-150261444 on Build GRCh38

View Full Product Details