Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 283934-284171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf96
Entrez Gene ID
140680
Gene Name
chromosome 20 open reading frame 96
Gene Aliases
dJ1103G7.2
UniGene
Hs.348112
Gene Chromosome Location
Chr.20: 270863-290778 on Build GRCh38
Transcripts
NM_080571.1 NM_153269.2 XM_006723543.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6402294 c.108G>A p.M36I Chr.20: 283987-283987 on Build GRCh38
1130671 c.209C>T p.T70M Chr.20: 284060-284060 on Build GRCh38
5346602 c.93C>T p.F31F Chr.20: 284071-284071 on Build GRCh38
3939287 c.191G>A p.W64* Chr.20: 283973-283973 on Build GRCh38
1025440 c.244G>T p.E82* Chr.20: 284025-284025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959900859 Chr.20: 284061-284061 on Build GRCh38
rs775131817 Chr.20: 283972-283972 on Build GRCh38
rs137926498 Chr.20: 283987-283987 on Build GRCh38
rs779936560 Chr.20: 284133-284133 on Build GRCh38
rs758364042 Chr.20: 284037-284037 on Build GRCh38
rs186161988 Chr.20: 284147-284147 on Build GRCh38
rs754123949 Chr.20: 284021-284021 on Build GRCh38
rs566102622 Chr.20: 283960-283960 on Build GRCh38
rs750846591 Chr.20: 284009-284009 on Build GRCh38
rs777199985 Chr.20: 284105-284105 on Build GRCh38
rs773918581 Chr.20: 283991-283991 on Build GRCh38
rs766975991 Chr.20: 284087-284087 on Build GRCh38
rs755306700 Chr.20: 284022-284022 on Build GRCh38
rs768117572 Chr.20: 284004-284004 on Build GRCh38
rs199714611 Chr.20: 283954-283954 on Build GRCh38
rs761028658 Chr.20: 284086-284086 on Build GRCh38
rs376791854 Chr.20: 284079-284079 on Build GRCh38
rs1003978704 Chr.20: 284033-284033 on Build GRCh38
rs6046931 Chr.20: 284123-284123 on Build GRCh38
rs1012464000 Chr.20: 284063-284063 on Build GRCh38
rs752826651 Chr.20: 284117-284117 on Build GRCh38
rs766782570 Chr.20: 284018-284018 on Build GRCh38
rs761198522 Chr.20: 283995-283995 on Build GRCh38
rs367585129 Chr.20: 284038-284038 on Build GRCh38
rs758512135 Chr.20: 284118-284118 on Build GRCh38
rs975923725 Chr.20: 284128-284128 on Build GRCh38
rs371871076 Chr.20: 284010-284010 on Build GRCh38
rs372813606 Chr.20: 284039-284039 on Build GRCh38
rs770965459 Chr.20: 283958-283958 on Build GRCh38
rs769438414 Chr.20: 283969-283969 on Build GRCh38
rs571813775 Chr.20: 284043-284043 on Build GRCh38
rs779269360 Chr.20: 284024-284024 on Build GRCh38
rs557519609 Chr.20: 284121-284121 on Build GRCh38
rs773771306 Chr.20: 284084-284084 on Build GRCh38
rs537291696 Chr.20: 284115-284115 on Build GRCh38
rs188822360 Chr.20: 283962-283962 on Build GRCh38
rs755918979 Chr.20: 284042-284042 on Build GRCh38
rs749079061 Chr.20: 284059-284059 on Build GRCh38
rs553428108 Chr.20: 284124-284124 on Build GRCh38
rs768337298 Chr.20: 284060-284060 on Build GRCh38
rs143380856 Chr.20: 284074-284074 on Build GRCh38
rs906929147 Chr.20: 284029-284029 on Build GRCh38
rs376594842 Chr.20: 284017-284017 on Build GRCh38
rs752858013 Chr.20: 284036-284036 on Build GRCh38
rs762536700 Chr.20: 283985-283985 on Build GRCh38

View Full Product Details