Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44172803-44173038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN11
Entrez Gene ID
11131
Gene Name
calpain 11
Gene Aliases
calpain11
UniGene
Hs.225953
Gene Chromosome Location
Chr.6: 44158811-44184402 on Build GRCh38
Transcripts
NM_007058.3 XM_006714984.1 XM_006714985.1 XM_006714987.1 XM_011514274.1 XM_011514275.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3352286 c.535C>T p.Q179* Chr.6: 44172946-44172946 on Build GRCh38
1079693 c.529-1G>T p.? Chr.6: 44172939-44172939 on Build GRCh38
259925 c.585G>T p.K195N Chr.6: 44172996-44172996 on Build GRCh38
7359416 c.619C>T p.R207C Chr.6: 44173030-44173030 on Build GRCh38
3352288 c.564A>G p.V188V Chr.6: 44172975-44172975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371708748 Chr.6: 44172920-44172920 on Build GRCh38
rs550444897 Chr.6: 44172913-44172913 on Build GRCh38
rs746493286 Chr.6: 44172902-44172902 on Build GRCh38
rs369057032 Chr.6: 44172840-44172840 on Build GRCh38
rs1014312106 Chr.6: 44172924-44172927 on Build GRCh38
rs780734751 Chr.6: 44172911-44172911 on Build GRCh38
rs532130406 Chr.6: 44172862-44172862 on Build GRCh38
rs754429712 Chr.6: 44172968-44172968 on Build GRCh38
rs374999083 Chr.6: 44173013-44173013 on Build GRCh38
rs372998163 Chr.6: 44172982-44172982 on Build GRCh38
rs772252566 Chr.6: 44172919-44172919 on Build GRCh38
rs201086941 Chr.6: 44172974-44172974 on Build GRCh38
rs372783271 Chr.6: 44172967-44172967 on Build GRCh38
rs368767278 Chr.6: 44172942-44172942 on Build GRCh38
rs751213228 Chr.6: 44172961-44172961 on Build GRCh38
rs772380025 Chr.6: 44172981-44172981 on Build GRCh38
rs760767972 Chr.6: 44172922-44172922 on Build GRCh38
rs776643350 Chr.6: 44172924-44172924 on Build GRCh38
rs367851443 Chr.6: 44172904-44172904 on Build GRCh38
rs765144796 Chr.6: 44172934-44172934 on Build GRCh38
rs562612246 Chr.6: 44172918-44172918 on Build GRCh38
rs902175705 Chr.6: 44172890-44172890 on Build GRCh38
rs926822044 Chr.6: 44172868-44172868 on Build GRCh38
rs375753243 Chr.6: 44172829-44172829 on Build GRCh38
rs757468399 Chr.6: 44172863-44172863 on Build GRCh38
rs752401365 Chr.6: 44172939-44172939 on Build GRCh38
rs879104924 Chr.6: 44172839-44172839 on Build GRCh38
rs938268871 Chr.6: 44172885-44172885 on Build GRCh38
rs763857652 Chr.6: 44172958-44172958 on Build GRCh38
rs369819193 Chr.6: 44172978-44172978 on Build GRCh38
rs747176501 Chr.6: 44172983-44172983 on Build GRCh38
rs755438819 Chr.6: 44172969-44172969 on Build GRCh38
rs776567421 Chr.6: 44173009-44173009 on Build GRCh38
rs62401124 Chr.6: 44172843-44172843 on Build GRCh38
rs749694993 Chr.6: 44172912-44172912 on Build GRCh38
rs759377540 Chr.6: 44172931-44172931 on Build GRCh38
rs754376153 Chr.6: 44172933-44172933 on Build GRCh38
rs375296253 Chr.6: 44172896-44172896 on Build GRCh38

View Full Product Details