Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15238696-15238969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD4
Entrez Gene ID
23476
Gene Name
bromodomain containing 4
Gene Aliases
CAP, HUNK1, HUNKI, MCAP
UniGene
Hs.187763
Gene Chromosome Location
Chr.19: 15236836-15332543 on Build GRCh38
Transcripts
NM_058243.2 XM_011527854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6287259 c.3811C>A p.Q1271K Chr.19: 15238952-15238952 on Build GRCh38
1680756 c.4016A>G p.E1339G Chr.19: 15238747-15238747 on Build GRCh38
6944280 c.3890A>C p.Q1297P Chr.19: 15238873-15238873 on Build GRCh38
3937909 c.3957C>T p.S1319S Chr.19: 15238806-15238806 on Build GRCh38
6968123 c.3958A>T p.M1320L Chr.19: 15238805-15238805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937066745 Chr.19: 15238899-15238899 on Build GRCh38
rs762603057 Chr.19: 15238863-15238863 on Build GRCh38
rs772586063 Chr.19: 15238884-15238884 on Build GRCh38
rs756018641 Chr.19: 15238810-15238810 on Build GRCh38
rs750347317 Chr.19: 15238919-15238919 on Build GRCh38
rs916929419 Chr.19: 15238918-15238918 on Build GRCh38
rs369387377 Chr.19: 15238829-15238829 on Build GRCh38
rs755237120 Chr.19: 15238925-15238925 on Build GRCh38
rs544008810 Chr.19: 15238847-15238847 on Build GRCh38
rs780701875 Chr.19: 15238869-15238870 on Build GRCh38
rs755041038 Chr.19: 15238838-15238838 on Build GRCh38
rs973091914 Chr.19: 15238841-15238841 on Build GRCh38
rs572472997 Chr.19: 15238721-15238721 on Build GRCh38
rs773613733 Chr.19: 15238753-15238753 on Build GRCh38
rs754054879 Chr.19: 15238922-15238922 on Build GRCh38
rs773962633 Chr.19: 15238884-15238885 on Build GRCh38
rs992519214 Chr.19: 15238917-15238917 on Build GRCh38
rs376762908 Chr.19: 15238779-15238779 on Build GRCh38
rs200486675 Chr.19: 15238881-15238881 on Build GRCh38
rs113881069 Chr.19: 15238953-15238953 on Build GRCh38
rs748234502 Chr.19: 15238844-15238844 on Build GRCh38
rs369815142 Chr.19: 15238926-15238926 on Build GRCh38
rs761012395 Chr.19: 15238761-15238761 on Build GRCh38
rs745340727 Chr.19: 15238870-15238872 on Build GRCh38
rs1019342567 Chr.19: 15238830-15238830 on Build GRCh38
rs371743668 Chr.19: 15238797-15238797 on Build GRCh38
rs758772498 Chr.19: 15238870-15238899 on Build GRCh38
rs773904017 Chr.19: 15238880-15238880 on Build GRCh38
rs772267056 Chr.19: 15238750-15238750 on Build GRCh38
rs546485417 Chr.19: 15238854-15238854 on Build GRCh38
rs763540393 Chr.19: 15238868-15238868 on Build GRCh38
rs868319763 Chr.19: 15238945-15238945 on Build GRCh38
rs749091959 Chr.19: 15238879-15238884 on Build GRCh38
rs376523850 Chr.19: 15238805-15238805 on Build GRCh38
rs202242420 Chr.19: 15238869-15238869 on Build GRCh38
rs763627798 Chr.19: 15238791-15238791 on Build GRCh38
rs761265784 Chr.19: 15238887-15238887 on Build GRCh38
rs375123750 Chr.19: 15238845-15238845 on Build GRCh38
rs112833044 Chr.19: 15238913-15238913 on Build GRCh38
rs200578182 Chr.19: 15238734-15238734 on Build GRCh38
rs769472173 Chr.19: 15238870-15238875 on Build GRCh38
rs139314630 Chr.19: 15238743-15238743 on Build GRCh38
rs780373163 Chr.19: 15238834-15238834 on Build GRCh38
rs764840421 Chr.19: 15238764-15238764 on Build GRCh38
rs112476530 Chr.19: 15238905-15238905 on Build GRCh38
rs772098711 Chr.19: 15238894-15238895 on Build GRCh38
rs760505424 Chr.19: 15238920-15238920 on Build GRCh38
rs1052018084 Chr.19: 15238858-15238858 on Build GRCh38
rs779357037 Chr.19: 15238845-15238846 on Build GRCh38
rs201443971 Chr.19: 15238785-15238785 on Build GRCh38
rs768491171 Chr.19: 15238881-15238882 on Build GRCh38
rs751390708 Chr.19: 15238794-15238794 on Build GRCh38
rs899942913 Chr.19: 15238732-15238732 on Build GRCh38
rs61740415 Chr.19: 15238836-15238836 on Build GRCh38
rs200976853 Chr.19: 15238855-15238855 on Build GRCh38
rs768878651 Chr.19: 15238731-15238731 on Build GRCh38
rs780461075 Chr.19: 15238720-15238720 on Build GRCh38
rs201420374 Chr.19: 15238949-15238949 on Build GRCh38
rs779654666 Chr.19: 15238871-15238894 on Build GRCh38
rs753254339 Chr.19: 15238864-15238878 on Build GRCh38
rs763138973 Chr.19: 15238812-15238812 on Build GRCh38
rs775086724 Chr.19: 15238857-15238857 on Build GRCh38
rs199802230 Chr.19: 15238921-15238921 on Build GRCh38
rs767464916 Chr.19: 15238895-15238895 on Build GRCh38
rs200422251 Chr.19: 15238941-15238941 on Build GRCh38
rs748141481 Chr.19: 15238895-15238897 on Build GRCh38
rs1000363868 Chr.19: 15238736-15238736 on Build GRCh38
rs760816131 Chr.19: 15238928-15238930 on Build GRCh38
rs755357952 Chr.19: 15238719-15238720 on Build GRCh38

View Full Product Details